Name derivation: Streptomyces phaeochro
Recognition and Cutting site: G CATG^C
C^GTAC G
Resulting DNA Ends: 3′ protruding ends
Prototype: SphI 
Isoschizomers: PaeI
Sensitivity to methylation:  EcoBI blocked if overlapped
Neoschizomers None
Anza RE generating compatible ends Anza™ 56 Hin1II, Anza™ 98 XceI
SphI nucleotide recognition sequence Figure color legend
References