High-throughput blood processing from 6 to 16 x 500mL blood bag capacity, matching your production needs
Select higher capacity rotors or match existing workflows with a choice of 4 Thermo Scientific™ HAEMAFlex™ rotors available for the new Thermo Scientific blood banking centrifuges.  Rotor buckets with Thermo Scientific™ Dura-Coat™ nickel-coating are also available, providing protection against corrosion.
Popular HAEMAFlex rotors
HAEMAFlex 16 Swinging Bucket Rotor
HAEMAFlex 12 Swinging Bucket Rotor
HAEMAFlex 6 Swinging Bucket Rotor
HAEMAFlex swinging bucket rotors
Unique 16 x 500mL blood bag capacity maximizes productivity for high-throughput blood processing needs.
Maximize total blood bags per spin with 12 x 500mL capacity up to 7,187 x g
Blood processing up to 8 x 550mL blood bags per run with performance of 4,600 rpm or 7,144 x g.
Address your lab’s regular- to medium-throughput blood processing needs with 6 x 550mL blood bag capacity at speeds up to 5,000 rpm.
Make working with large volumes easier and more convenient using the protocols you've already validated. Our large capacity centrifuges for blood banking provide enhanced ergonomics and compact design plus quick set-up of traceable, reproducible runs.
Featured blood banking centrifuges videos
Resources
Support
  • Unity Lab Services 
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.