Pathogen Detection Research by Real-Time PCR
Your trusted partner in pathogen detection research
When it comes to pathogen detection, quantitative polymerase chain reaction (qPCR) is the most established and successfully implemented molecular technique to date. It can detect slow-growing, difficult-to-cultivate, or uncultivatable microorganisms, and can be used when traditional microbiological techniques are inadequate, ambiguous, time-consuming, difficult, and costly.
As a leader in real-time PCR, Thermo Fisher Scientific offers a complete solution for pathogen detection research—from real-time PCR systems to master mixes and TaqMan™ Assays, along with services and support to help you validate your pathogen detection workflow. The choice is yours, whatever your needs.
Molecular biological methods for the detection and characterization of microorganisms have revolutionized investigative microbiology and are now part of routine specimen processing. PCR techniques have led the way into this new era by allowing rapid detection of microorganisms that were previously difficult or impossible to detect by traditional microbiological methods. In addition to detection of fastidious microorganisms, more rapid detection by molecular methods is now possible for pathogens of public health importance. Molecular methods have now progressed beyond identification to detect antimicrobial resistance genes and provide public health information such as strain characterization by genotyping. Understanding of certain microorganisms has been improved by viral resistance detection and viral load testing for the monitoring of responses to antiviral exposure. With the advent of multiplex PCR, real-time PCR, and improvements in efficiency through automation, the costs of molecular methods are decreasing such that the role and use of molecular methods will further increase.
Quantitative polymerase chain reaction (qPCR) can detect slow-growing, difficult-to-cultivate, or uncultivatable microorganisms, and can be used when traditional microbiological techniques are inadequate, ambiguous, time-consuming, difficult, and costly.
Inherent technical limitations of PCR are present, but they are reduced in laboratories that use standardized protocols, conduct rigid validation protocols, and adhere to appropriate quality-control procedures. Improvements in PCR, especially probe-based real-time PCR, have broadened its molecular detection capabilities in pathogen detection to complement and even surpass traditional methods in some situations. 
Explore our complete list of pathogen detection assays:
Complete list of TaqMan pathogen detection assays
Order now
The freedom of flexibility: choose a format or solution that works for you
TaqMan chemistry is at the core of our pathogen detection assays. TaqMan Assays are the gold standard chemistry for real-time PCR testing, and provide the accuracy and sensitivity for enhanced performance that is required for pathogen detection.
Explore our formats:
Explore our complete solutions:
  • Outbreak pathogen detection: Zika virus
  • TaqMan™ Vaginal Microbiota detection assays—coming soon
  • TaqMan™ antimicrobial resistance assays—coming soon
  • TaqMan™ gastrointestinal microbial detection assays—coming soon
  • TaqMan™ respiratory microbial detection assays—coming soon
Thermo Fisher Scientific's response to Zika virus
In May 2015, the Pan American Health Organization (PAHO) issued an alert regarding the first confirmed Zika virus infection in Brazil, and on February 1, 2016, the World Health Organization declared Zika virus a public health emergency of international concern. In response, Thermo Fisher Scientific has worked diligently to develop a lyophilized one-step real-time PCR multiplex TaqMan Assay for molecular detection of RNA from the dengue, chikungunya, and Zika viruses.
Read more
Instruments and master mixes for qPCR pathogen detection research
As a leader in PCR innovation, we offer you the gold standard in real-time PCR instrumentation. With our Applied Biosystems™ real-time PCR platforms, you get true value with excellent performance, reliability, and world-class support. Designed for compatibility with TaqMan Assays and the flexibility of unlimited real-time chemistry choices, we make it easy for you to get started with your experiments. The QuantStudio™ family of instruments enables you to obtain the results you need, connect and collaborate with colleagues, and achieve your research goals.
TaqMan™ Fast Virus 1-Step Master Mix is designed for reliable, high-sensitivity real-time RT-PCR even in the presence of common reaction inhibitors. The features of the kit have been selected to enhance virus detection on commonly used sample types. A single-tube format allows for uniform handling and processing for both RNA and DNA viruses.
Technical support and compliance services
Do you need technical support or training? We can help.
Find out more
Depending on your regulatory requirements, services may be necessary to comply with CAP, CLIA, ISO, GMP, USDA, and other standards.  All our compliance services come with audit-ready documentation.
Find out more
tech support in the lab