Variable nucleotide tandem repeats (VNTRs) are repeating sequences of multi-base segments of DNA. If the repeat is equal to or less than 6 bases, NTRs are named microsatellites, also known as short tandem repeats (STRs). One common example of a microsatellite is a (CA)n repeat, where n varies between alleles. Because microsatellites are polymorphic DNA loci present throughout the genome, microsatellite genotyping is a widely accepted tool for a variety of research applications such as linkage mapping studies, association studies, and identification of organisms.
Our five-dye chemistry increases the number of microsatellite markers that can be run in a single capillary, substantially improving the throughput.
Step-by-Step Guide to Microsatellite STR Analysis
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify the DNA region containing the microsatellite of interest.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
- Genotyping S S S S S S S
- Analysis for Forensics/HID N N S S S S S
- Instability/RER A A A A A A A
Analyze the RFLP profile.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify the DNA region containing the microsatellite of interest.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
- Genotyping S S S S S S S
- Analysis for Forensics/HID N N S S S S S
- Instability/RER A A A A A A A
Analyze the RFLP profile.
Which Data Analysis Software Is Right for You?
Step-by-Step Guide to Microsatellite STR Analysis
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify the DNA region containing the microsatellite of interest.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
- Genotyping S S S S S S S
- Analysis for Forensics/HID N N S S S S S
- Instability/RER A A A A A A A
Analyze the RFLP profile.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify the DNA region containing the microsatellite of interest.
Recommended Products: Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
- Genotyping S S S S S S S
- Analysis for Forensics/HID N N S S S S S
- Instability/RER A A A A A A A
Analyze the RFLP profile.
Which Data Analysis Software Is Right for You?
Highlighted Products for Microsatellite STR Analysis:
3500 Series Genetic Analyzers
GeneMapper® Software
Verti Thermal Cycler
Visistat reference component
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.
Fragment Analysis Software Support Center