Share |
Quicklinks
Related Product Information
Go to...
Calculation worksheets for LanthaScreen® Kinase Activity Assay
Kinase for EC80 determination:
Kinase:   ____ Stock = ____ mg/mL = _____ μg/mL
1X = _____ μg/mL (from validation packet)
2X = ____   μg/mL
To prepare 250 μL:
V1          x         C1        =         V2         x             C2
                      [Initial]                                           [Final 2X]
Kinase
V1          x       μg/mL    =      250 μL     x              μg/mL
V1 = μL
If V1 is less than 2 μL, perform a 10-fold dilution by adding 2 μL of kinase to 18 μL of 1X  Kinase Buffer A and add 10 x V1.
Buffer:
250 μL – __________μL = ________μL 1X Kinase Buffer A
Add the calculated volume of kinase to the appropriate volume of 1X Kinase Buffer A. Keep the diluted kinase on ice until needed.
Note:
If your kinase requires additives such as calcium and calmodulin, please be sure to substitute a complete kinase reaction buffer for 1X Kinase Buffer A.
2X Substrate/2X ATP:
You may need to convert mg/mL listed on the tube to nM. The molecular weight of the substrate is listed on the COA.
Substrate____
1X = ____ nM
2X = ____  nM
ATP Stock = 10 mM = 10,000 μM
1X (ATP K m) = _____μM (from validation packet)
2X = _____ μM
To prepare  ______ μL:
V1                x          C1              =            V2              x             C2
                           [Initial]                                                    [Final 2X]
Substrate V1                x         _____nM         =      ____  μL       x          _____nM
V1  = _____ μL
ATP V1                x          10,000 μM    =   _____μL        x                _____μM
V1  = _____ μL
If V1 is less than 2 μL, perform a 10-fold dilution by adding 2 μL of 1X kinase to 18 μL of Kinase Buffer A and add 10 x V1.
Buffer:  ____μL – ____μL – ____μL = ____μL 1X Kinase Buffer A
Add the calculated volume of substrate and ATP to the appropriate volume of 1X Kinase Buffer A.
2X EDTA/2X Antibody:
EDTA Stock = 500 mM
1X = 10 mM
2X = 20 mM
Important: Centrifuge the antibody tube at approximately 10,000 x g for 10 minutes and pipet desired volume from the top of the solution.
Antibody Stock = ____ μM = ____ nM
1X = 2 nM
2X = 4 nM
To prepare ____ μL:
V1              x                     C1                =             V2            x                  C2
                                      [Initial]                                                          [Final2X]
EDTA  V1             x                500 mM            =       ____ μL        x               20 mM
V1 =  ____μL                                                                                                    
Antibody V1              x                ____nM            =       ____μL         x                  4 nM
V1 =  ____μL   
Buffer:    ____μL – ____μL – μL = μL TR-FRET dilution buffer
Add the calculated volume of EDTA and antibody to the calculated volume of TR-FRET
dilution buffer.
4X (EC80) Kinase for IC50 determinations:
Kinase: Stock = ng/mL (from Step 1.2)
1X (EC80) = ng/mL (from Step 1.10)
4X (EC80) = ng/mL
To prepare μL:
V1          x         C1        =         V2         x             C2
                      [Initial]                                             [Final 4X]
Kinase
V1          x     ___ng/mL    =      1,000 μL     x              μg/mL
V1 = μL
Buffer: 250 μL – ______  μL = ______  μL   1X Kinase Buffer A
Add the calculated volume of kinase to the appropriate volume of 1X Kinase Buffer A.
2X Substrate/2X ATP:
You may need to convert mg/mL listed on the tube to nM. The molecular weight of the substrate is listed on the COA.
Substrate_____
1X = ____ nM
2X = ____ nM
ATP Stock = 10 mM = 10,000 μM
1X (ATP Km) = ____ μM (from validation packet)
2X = _____  μM
To prepare ______ μL:
V1                x          C1              =            V2               x             C2
                             [Initial]                                                        [Final 2X]
Substrate V1                x           _____nM         =   ____  μL        x                  _____nM
V1  = _____ μL
ATP V1                x          10,000 μM    =      _____μL         x                  _____μM
V1  = _____ μL
If V1 is less than 2 μL, perform a 10-fold dilution by adding 2 μL of kinase to 18 μL of 1X Kinase Buffer A and add 10 x V1.
Buffer:   _____
μL – _____μL   –  _____  μL = _____ μL 1X Kinase Buffer A
Add the calculated volume of substrate and ATP to the appropriate volume of 1X Kinase Buffer A
2X EDTA/2X Antibody:
EDTA Stock = 500 mM
1X = 10 mM
2X = 20 mM
Antibody Stock = μM = nM
1X = 2 nM
2X = 4 nM
To prepare μL:
V1                x          C1              =            V2               x             C2
                           [Initial]                                                        [Final 2X]
EDTA V1                x         500 nM         =     ____  μL        x                20 nM
V1  = _____ μL
Antibpdy V1                x          ____ μM    =      _____μL         x                  4 nM
V1  = _____ μL
Buffer:  
____ _ μL – _____ μL – _____ μL =  ____ μL TR-FRET dilution buffer
Add the calculated volume of EDTA and antibody to the calculated volume of TR-FRET dilution buffer.
TOP
Go to...
LT129       1-Jan-2010