Easy to read. Simple to perform
Easy to read. Simple to perform
Combine the simplicity and ease of use of the diffusion method with the accuracy of an Minimum Inhibitory Concentration (MIC) test. Firm but flexible, Thermo Scientific™ M.I.C. Evaluator™ (M.I.C.E.™) strips are easy to handle and apply to pre-inoculated agar plates. Upon application, the antimicrobial is released from the strip, forming a clear, easy-to-read zone.
Maintain performance integrity until you're ready to test and ensure accurate results. Each easy-to-handle strip is individually foil wrapped with desiccant to maintain quality until the point of use.
Related products
Join the fight against antibiotic resistance
Learn about the importance of antimicrobial stewardship policies and practices and effective implementation. 
 View slides and certificate
Professor Dilip Nathwani answers your questions