Assay specifications
  Limit of detection   ≤ 10,000 copies/assay wella
  Limit of quantitation   ≤ 20,000 copies/assay wellb
  Linear dynamic range   ≥ 2.5 logsc
  Assay CV   ≤ 15% intra-assay;≤ 20% inter-assay
  Compatible sample types   Cultured cells, bacteria, whole blood, 
  PAXgene blood or dried blood spots, 
  fresh/frozen tissues(human, animal or plant), 
  FFPE samples, purified DNA
  Assay format   96-well plated
  Targets/well   2-80
  Automation compatible
  Yese
Defined as signal greater than background plus three standard deviations of the background.
Defined as the signal just above the lowest signal that obtains an 80–120% spike recovery.
Defined as the assay window that consistently achieves an 80–120% accuracy of fold change.
Contact technical support for information on a 384-well version.
Batch or full automation using standard automation equipment. Contact technical support for protocol details and assistance in setting up your automation system.
Multiplex DNA copy number analysis
The QuantiGene Plex DNA Assay allows simultaneous detection of multiple copy number alterations in a single well. In the example below, an eight-plex DNA assay was used to quantify Her2 and other genes in SKBR3 breast cancer cells and control cell lines (normal skin fibroblasts and MDA-231 cells) (Figure 1). The QuantiGene Plex DNA Assay showed a ~7-fold amplification of the Her2 gene and two other genes on chromosome 17, and was concordant with that of a bDNA Her2 DNA fluorescent  in situ hybridization (FISH) assay (Figure 2), where a 7-fold amplification of Her2 was detected in SKBR3 cells relative to control HeLa cells.
Figure 1: QuantiGene Plex DNA Assay in SKBR3, normal skin fibroblast, and MDA-231 cells. The ratio of SKBR3 to control copies was ~7. Two other genes on chromosome 17 were amplified and one gene was not. Control genes on chromosomes 1, 5, and 8 in SKBR3 breast cancer cells and control cell lines did not show any DNA copy number amplification.
Figure 2: FISH DNA assay showing Her2 amplified in SKBR3 cells relative to HeLa cells. The ratio of SKBR3 to HeLa copies was ~7. Note: The FISH DNA assay is not currently commercially available.
Same results from any starting material
The QuantiGene Plex DNA Assay offers reproducible and reliable results from a variety of starting materials. In this example, purified genomic DNA as well as lysates from seed and leaves were prepared from the same transgenic lettuce plant. The samples were assessed using the QuantiGene Plex DNA Assay with a panel of 10 targets. After normalization to gene 10, the results indicated the same ratio of gene copy number for each of these genes in all three samples.
Figure 3: Results of the QuantiGene Plex DNA Assay with different sample sources, including purified genomic DNA (gDNA) as well as lysates from seeds (seed) and leaves (leaf) of a transgenic lettuce plant. Normalized data with gene 10 are shown to indicate the relative DNA ratio.
Drug metabolism panel – DNA copy number
The QuantiGene Plex DNA Assay offers single-copy number discrimination for accurate classification of copy number variation in drug metabolism studies. The assay can be used as a companion to Affymetrix DMET™ Arrays or to validate array data. In this case, a nine plex assay was used to quantify six target genes, SULT1A1, GSTM1, CYP2D6, CYP2A6, GSTT1, and UGT2B17, normalized using three genes (RPPH1, SNAP25, and PMP22) in 20 Coriell samples (nine Asian, six Yoruba, and five Caucasians) and one ParagonDx sample (Caucasian). The results were analyzed using the background-subtracted average MFI value of target and control genes. The target genes’ MFI value was normalized by the geometric mean of the three control genes’ MFI. The control genes are considered to be N=1. In the data shown, F: female; M: male; J/M: Japanese male; J/F: Japanese female; C/M: Chinese male; and C/F: Chinese female.
Figure 4: Ethnic group analysis of SULT1A1 DNA copy number using the QuantiGene Plex DNA Assay.
Figure 5: Ethnic group analysis of CYP2D6 DNA copy number using the QuantiGene Plex DNA Assay.
Figure 6: Ethnic group analysis of UGT2B17 DNA copy number using the QuantiGene Plex DNA Assay