QuantiGene Plex DNA Assay workflow
The QuantiGene Plex DNA Assay is a hybridization-based assay using the xMAP Luminex magnetic beads and performed on 96-well plates. The assay is based on DIRECT quantification of the DNA targets using xMAP Luminex beads for multiplexing of 3 to 33 DNA targets and branched DNA (bDNA) signal amplification technology. On the first day the sample is lysed to release the DNAs, and incubated overnight with target-specific probe sets panel. On the second day the signal amplification tree is built via sequential hybridization of PreAmplifier (PreAmp), Amplifier (Amp), and Label Probe (LP). Each amplification unit gives a 400x signal amplification and there are six amplification units per target DNA copy, leading to a 2,400x signal amplification per copy DNA. The signal is detected by adding SAPE substrate and using a Luminex instrument for the readout.
QuantiGene Plex DNA Assay workflow
Step 1. Sample Preparation: Samples are lysed to release and stabilize DNAs. The DNA assay works with a variety of samples such as: cultured cells, human, plant and animal tissues, FFPE tissues, whole blood and PAXGene blood, or purified DNA. 
Step 2. Target Hybridization: Overnight hybridization in the 96-well plates with the target specific probe sets panel (Capture extenders – CE’s, Label Extenders – LE’s, and blocking probes).  
Step 3. Signal Amplification: Signal amplification is achieved using branch DNA (bDNA) technology. A Pre-Amplifier (PreAmp) molecule hybridizes to each pair of Label Extenders, but not to individual probes. Then, multiple Amplifier (Amp) molecules hybridize to each PreAmp. Finally, multiple Label Probe oligonucleotides hybridize to each Amp. 
Step 4. Detection: Addition of streptavidin phycoerythrin (SAPE) generates a signal that is proportional with the amount of target DNA present in the sample. The signal is read using a Luminex instrument.
Video: How it works