Designed for liquid phase PCR or immuno-PCR
ThermoScientific™ Nunc™ TopYield™ Breakable Modules are specifically designed for liquid phase PCR or immuno-PCR. The unique design offers error-free flexibility, and each well locks in the frame and maintains the same height to provide accurate reading and efficient washing.
  • Made of polycarbonate
  • Consists of 1 x 8 strips to be fitted into a 96-well format frame
  • V-shaped wells with flat, readable bottoms and thin walls (0.35 mm)
  • Excellent protein binding
  • Excellent heat transmission
  • Comparable with 0.2 mL PCR tube
  • Compatible with most 0.2 mL thermal cycler formats
Your dedicated partner for In Vitro Diagnostics