AutoRELI™ 48 Instrument (Product Code 899.05)
A fully automated probe hybridisation and strip detection system
  • An efficient run time - Only 2hrs 20mins for 48 samples
  • Identical protocol for class I and class II loci - Test multiple loci during one run
  • Process flexibility - process 2 to 48 samples per run
  • Accurately controlled heating system - Uniform and precise temperature control across the 48 wells
  • Improved waste collection - Reducing volume of toxic waste
  • For invitro diagnostic use
RELI-Scan™ Instrument (Product Code 899.06)
  • RELI-Scan InstrumentGet typing results directly from the AutoRELI™ 48 tray
  • Eliminates Manual Handling of Strips - Strips scanned in AutoRELI™ 48 tray
  • One step scanning process - 48 results from a single scan
  • Results interpreted as strips are scanned
  • Saves time, increases throughput - 48 strips scanned in 12 minutes
  • For research use only, not intended for diagnostic or therapeutic use