A solid tumor next-generation sequencing assay for biomarker analysis
The Oncomine™ Focus Assay is a targeted, multi-biomarker assay that enables you to target hotspots, SNVs, indels, CNVs, and gene fusions from DNA and RNA in a single workflow.
Designed for clinical and translational cancer research, the Oncomine Focus Assay leverages Ion AmpliSeq™ technology to enable robust results, low input amounts, and analysis of low quality samples, and is compatible with FFPE fine needle aspirates and core needle biopsies.
Request info       Download analytical performance white paper ›
Content
  • Simultaneous detection of hundreds of variants across 52 genes relevant to solid tumors
  • Targets relevant hotspots, SNVs, indels, CNVs, and gene fusions from DNA or RNA in a single workflow
  • Includes solid tumor genes targeted by on-market oncology drugs and published evidence
  • Content driven by the Oncomine™ Knowledgebase, which includes content aligned to approved therapies and indications
Oncomine Focus Assay gene list
ABL1  ERBB2  GNAQ  MYC 
AKT1  ERBB4  HRAS  MYCN 
AKT3  ERBB3  IDH1  NRAS 
ALK  ERG  IDH2  NTRK1 
AR  ESR1  JAK1  NTRK2 
AXL  ETV1  JAK2  NTRK3 
BRAF  ETV4  JAK3  PDGFRA 
CCND1  ETV5  KIT  PIK3CA 
CDK4  FGFR1  KRAS  PPARG 
CDK6  FGFR2  MAP2K1  RAF1 
CTNNB1  FGFR3  MAP2K2  RET 
DDR2  FGFR4  MET  ROS1 
EGFR  GNA11  MTOR  SMO 
Comparison of Oncomine assay and Ion AmpliSeq panel content
Targeted next-generation sequencing (NGS) is empowering laboratories of all sizes to confidently pioneer breakthroughs in cancer research. At the core of these breakthroughs are assays and panels designed to achieve highly sensitive results from minimal sample input—to help deliver information that may lead to the next companion diagnostic test or therapy in the future.
Clinical cancer research requires countless decisions. Of critical importance are the decisions you make about assay content. This comparison of the Oncomine™ assay and Ion AmpliSeq™ panel content is designed to help you tailor your content choices.
Examples of Oncomine assays and Ion AmpliSeq panels and a subset of their associated genes and tumor types for research use. The Oncomine Knowledgebase shows that the specific gene had a variant detected in specific tumor type at a minimum of a 1% frequency. Additional genes are available on these and other Oncomine assays and Ion AmpliSeq panels.
 Click to enlarge
Performance
  • Reproducible and sensitive detection of variants across multiple cancer types
  • Compatible with as little as 10 ng sample, including FFPE fine needle biopsies and core needle aspirates
  • Enables results from up to 6 samples and 2 controls per run for DNA and RNA in a single workflow
  • NGS results in days rather than weeks, leveraging Ion Torrent™ sequencing technology and the Ion S5™ System or the Ion PGM™ System
  • Includes Ion Select consumables and reagents manufactured to rigorous standards in our ISO 13485-compliant facility
Analytical performance summary
Across variant types, the Oncomine Focus Assay achieves high sensitivity, specificity, and reproducibility using clinical research samples sequenced on the Ion S5 System.
  Sensitivity PPV (Specificity) Reproducibility
SNVs 98.77% 99.63% 99.6%
Indels 100% 100% 100%
CNVs 96.42% 100% 96.42%
Gene fusion 100% 100% 100%
Overall 98.70% 99.67% 99.01%
Table 1. Sensitivity, specificity, and reproducibility across variant types using the Oncomine Focus Assay on the Ion S5 System.
Reporting
Features enabled with Oncomine informatics:
  • Prioritize variants, find the relevant few
  • Call variants, identify and annotate cancer drivers
  • Quickly create custom, lab-generated reports
  • Customizable report templates available in 10 languages
Workflow
The Oncomine NGS oncology workflow enables you find the relevant few cancer drivers, with a report including interpretation of the findings.
Request more information