Impact Weighers for Continuous Mass Flow Measurement
Improve product quality, reduce waste
Impact weighers continuously measure the mass flow rate and total mass of free-flowing particulate materials in mechanical conveying systems, without interrupting the flow of material.
Thermo Scientific™ Ramsey™ impact weighers provide a low-cost, highly accurate method for continuous mass flow measurement of dry solids and powders for inventory process control. They can help you save money by reducing waste and improving product quality, and are ideal for applications in vertical flow streams where weighbelt feeders or belt conveyor scales will not fit or cannot be applied.
Featured impact weighers
Achieve continuous mass flow measurement in belt conveying systems without interrupting the flow of material using the Ramsey DE10 Impact Weigher. The Ramsey DE10 is designed to measure bulk solids with flow rates from 4.54 t/h to 726 t/h (5.0 tn/h to 800 tn/h).
Measure the mass flow rate and total mass of free-flowing particulate materials with the Ramsey DE20 Impact Weigher. The Ramsey DE20 is designed to measure bulk solids with flow rates from 0.27 t/h to 36 t/h (0.3 tn/h to 40 tn/h).
Product weighing and totalization functions are performed with the Ramsey Micro-Tech 9106 Impact Weigher electronics, available in field-mount and panel-mount models. These electronic integrators process signals from the DE10 and DE20 impact weighers and display flow rate and total weight.
Invest in peace of mind with a Thermo Scientific service agreement. Ensure minimum downtime with known costs and priority response times. Let us tailor a plan to meet your needs. For parts, technical support, or field assistance, please call 800-445-3503.