Ion Chromatography Innovations
Innovating ion chromatography for today’s demands
Learn about our latest innovations in ion chromatography (IC). Designed with you in mind, they provide leading edge capabilities that help you meet the ever increasing requirements of productivity, performance and simplicity.
IC innovation features
Capabilities brochure
Learn how Reagent-Free™ IC (RFIC™) enables eluent generation by just adding water and other components that make RFIC systems work.
White paper
Explore the future of suppressor technoology – continuous operation, high-capacity, ease-of-use, backpressure tolerance, peak efficiency, and current efficiency.
High Pressure Ion Chromatography (HPIC)
With the ability to operate with ease at high backpressures, Thermo Scientific™ Dionex™ HPIC™ systems deliver a new level of speed and resolution.
Capillary Ion Chromatography
See what the world’s first Dionex HPIC capillary systems and columns have to offer: increased productivity from always-on and walk-up ready capillary IC,  with all the benefits of high pressure operation.
Charge Detection in Ion Chromatography
Charge detection is a membrane-based technology that detects ions in proportion to their charge and concentration, permitting reliable quantification of known and unknown compounds with a single standard.
Chromeleon CDS Software Resources
Here’s a collection of resources to learn about the features that make this software both powerful and easy to use.