SMART Digest and SMART Digest IA kits
SMART Digest and SMART Digest IA Kits
Fast, easy, reproducible, sensitive protein digestion and immunoaffinity capture
Use the latest advance in protein digestion for peptide mapping in biomarker and biotherapeutics characterization and quantitation. Thermo Scientific SMART Digest Kits provide amazingly fast, simple, reproducible, and sensitive protein digestion with immunoaffinity capture, giving you higher confidence in your chromatography and liquid chromatography-mass spectrometry (LC-MS) results.
Quick links
Innovation in protein sample preparation
Enjoy the SMART Digest difference in your peptide mapping and peptide quantitation data.
  • Easy 3-step protocol
  • Better reproducibility than in-solution digestion
  • Ability to automate with  Kingfisher platform
  • Much faster sample processing time (hours) compared to in-solution digestion (overnight)
  • Higher throughput and greater sensitivity
  • Higher confidence in your chromatography and LC-MS results
Featured SMART Digest videos
Ajinomoto Althea is a contract development and manufacturing company. This is their experience using SMART Digest kits.
Watch this introductory animation to learn how SMART Digest kits provide easy, fast, and highly reproducible protein digestion.
SMART Digest kits for peptide mapping and peptide quantitation
  • Choose from trypsin, chymotrypsin, or proteinase K kits for sequence-specific cleavage of your protein.
  • Use SMART Digest ImmunoAffinity (IA) kits for immunoaffinity capture and trypsin digestion in a single well. The Immunoaffinity reagent (streptavidin, protein A, or protein G) captures the protein and increases sensitivity by purifying low level proteins from complex biological matrices. The trypsin enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions.
SMART Digest Trypsin Kits
Uses trypsin to cleave the C-terminal side of lysine and arginine amino acid residues of peptides.
Order now
SMART Digest Chymotrypsin Kits
Uses chymotrypsin to cleave the peptide bonds formed by aromatic residues such as tyrosine, phenylalanine, and tryptophan.
Order now
SMART Digest Proteinase K Kits
Uses proteinase K kits to cleave the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups.
Order now
SMART Digest ImmunoAffinity (IA) Streptavidin Kits
The Streptavadin version of the kit allows for purification of various biomolecules via the formation of strong streptavidin-biotin bond.
Order now
SMART Digest ImmunoAffinity (IA) Protein A Kits
The Protein A version of the kit allows for targeted purification of immunoglobulins due to the strong affinity for binding to the Fc portion of the molecule.
Order now
SMART Digest ImmunoAffinity (IA) Protein G Kits
The Protein G version of the kit allows for targeted purification of immunoglobulins due to the strong affinity for binding to the Fab and Fc portion of the molecule.
Order now
Compare SMART Digest and in-solution techniques
Perform protein digestion in hours
SMART Digest vs. Conventional In-Solution Digest Protocol
Calculate how much time you will save
Just select the size of your protein to see how much time you will save using SMART Digest kits.
Improve reproducibility
Better reproducibility
Overlay of 13 consecutive chromatographic runs of a peptide sample separated on an analytical Thermo Scientific Acclaim 120 C18 column (RP18, 2.2 µm, 2./1 x 250 mm) and prepared from a mAb, digested with the SMART Digest kit.
Learn more
Easy 3-step protocol
Learn more
Perform protein digestion in hours
SMART Digest vs. Conventional In-Solution Digest Protocol
Calculate how much time you will save
Just select the size of your protein to see how much time you will save using SMART Digest kits.
Improve reproducibility
Better reproducibility
Overlay of 13 consecutive chromatographic runs of a peptide sample separated on an analytical Thermo Scientific Acclaim 120 C18 column (RP18, 2.2 µm, 2./1 x 250 mm) and prepared from a mAb, digested with the SMART Digest kit.
Learn more
Easy 3-step protocol
Learn more
Educational webinars
Attend SMART Digest webinars to learn more
  • Don’t miss a thing on your peptide mapping journey
  • Take the smart route to protein digestion
  • Sample preparation technologies for improved peptide quantitation workflow
  • Peptide separations with pinpoint precision
Register to view webinars
Resources
Product literature
Application notes
Frequently asked questions (FAQs)
Manuals