Characterize the full diversity of biomolecules
Characterize the full diversity of biomolecules
Get high resolution, high efficiency separations of biological samples. Our unique column chemistries for biological samples have been specifically designed for the separation of proteins, peptides, monoclonal antibodies, carbohydrates, nucleic acids, and more.
Popular Bio LC columns
ProPac™ WCX-10 LC Columns
EASY-Spray™ C18 LC Columns
DNAPac™ RP Columns
Bio LC column categories
High resolution and high efficiency set these columns apart. We offer a full range of affinity, ion exchange, reversed phase, hydrophobic interaction, and size exclusion columns for excellent analytical characterization of proteins.
These ion exchange columns are ideal for analyzing charged peptides in proteomic applications. If you need to resolve hundreds or even thousands of peptides, these reversed phase peptide columns will meet the challenge.
These HPLC columns are unique to the industry. Use them to separate neutral glycans, both neutral and charged glycans, or to purify and isolate carbohydrates.
Their ability to provide exceptional high resolution of nucleic acids (+1 and -1), including isomer separations, sets these columns apart from the rest.
These columns are specifically designed for monoclonal antibody (mAb) characterization, mAb titer analysis, and the separation of mAbs, mAb fragments, antibody-drug aggregates, and related biologics.
Featured Bio LC application videos
Monoclonal Antibody Analysis Update: Better Separations of MAb variants
Expert Ilze Birznieks introduces new tools for enabling cancer researchers as well as scientists developing biotherapeuitcs focused on monoclonal antibodies.
Glycan Analysis Update: Achieving Greater Resolution
Investigating glycans as disease markers and antibody-based biopharmaceuticals just got easier with the introduction of the Thermo Scientific™ GlycanPac™ AXR-1 HPLC column.
Featured LC / LC-MS webinar
Featured Webinar: Fast, Robust LC and LCMS Workflows for the Comprehensive Characterization of Bio-Therapeutic Proteins
Fast, robust LC and LC-MS workflows for the comprehensive characterization of biotherapeutic proteins
  • Orbitrap technology—a powerful protein characterization tool
  • New advances in analytical method development
  • 10 min runs with intelligent and integrated chromatography
  • A complete workflow for biopharma analysis
Are you using the right Bio LC column?
Access the latest version of our Chromatography Columns and Consumables Catalog in PDF and online formats. The enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Download catalog ›