Viper Fittings
Fingertight zero-dead volume fittings
Looking to make perfect LC connections without any extra tools? Use Thermo Scientific Viper and nanoViper Fingertight Fittings throughout the entire fluidic pathway of your LC system.
Viper and nanoViper connections allow you to quickly connect LC modules, valves, and columns for leak-free connections that improve chromatographic results by minimizing dead volume. Viper and nanoViper fittings require no tools for installation, and combine simplicity with high performance.
Quick links
Popular Viper fingertight fittings
Featured Viper fingertight fittings
Viper UHPLC fittings
Viper fingertight fittings seal at the tip of the tubing, virtually eliminating dead volumes and making it easier to plumb your HPLC or UHPLC system without any tools for reproducible leak-free connections to pressures up to 1500 bar.
Reusable and available in stainless steel (SST), PEEK and a biocompatible alloy (MP35N).
Find specific fittings and capillaries or purchase complete solution kits for your HPLC and UHPLC system or applications
nanoViper LC fittings
Avoid data-destroying dead volumes in the capillary connections of your nano LC system. Get consistently perfect connections for robust chromatography with nanoViper fingertight fittings.
Reusable nanoViper fittings consist of PEEK-shielded fused silica tubing and a fingertight sealing fitting.
Find specific fittings and capillaries or purchase complete solution kits for your nano LC system or applications:
Overview of Viper fingertight fitting system
  Viper
Stainless Steel
Viper
PEEK
Viper
MP35N
nanoViper
Connection principle
Fingertight fitting
Tubing type
Flexible
stainless steel
(SST)
PEEK
Biocompatible
MP35N
Biocompatible
Shielded fused silica + PEEK
Wetted material
PEEK, SST PEEK PEEK, MP35N PEEK, fused silica
Maximum pressure
1310 bar 345 bar 1500 bar 1200 bar
Maximum temperature
120°C 30°C 120°C 120°C
Viper nut threading
Compatible with 10-32 threaded ports
Outer diameter
0.79mm (1/32")
Inner diameter (ID)
100µm (0.004")
130µm (0.005")
180µm (0.007")
65µm (0.0025")
90µm (0.0035")
130µm (0.005")
100µm (0.004")
130µm (0.005")
180µm (0.007")
10µm (0.0004")
20µm (0.0008")
50µm (0.0020")
75µm (0.0030")
100µm (0.004")
150µm (0.006")
Available lengths
65mm to 950mm 65mm to 850mm 65mm to 950mm 70mm to 1100mm
*customizable lengths
Fittings kits
Order all of the fittings and capillaries for your application in a single kit.
These kits contain all the components to effortlessly plumb your
system from start to finish.
UHPLC Fittings Kits
nano LC Fittings Kits
= Not applicable = Poor = Good = Better = Best
Featured Viper videos
Chromatography columns and consumables catalog
Access the latest version of our Chromatography Columns and Consumables Catalog to ensure that you're optimizing your applications! Our enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Download catalog