Gino Tellier
Technical Support Specialist
Environmental and Process Monitoring
Meet our techs: Gino Tellier
Gino brings over 30 years of expertise in service, quality assurance, manufacturing, operations and technical support to our customers looking to optimize in ambient particulate applications.
Our techs are here to answer your questions, just email us.
Ask our techs ›
If you like this month's Q&A, check out our Talk Tech archive.
Question
Answer
For  Thermo Scientific™ 5014i Beta Continuous Ambient Particulate Monitor and the Model 5030i SHARP Monitor, it's good practice to perform leak checks every time you audit the unit’s performance. At the very least, the leak check should be performed once a month.  You don’t want to be collecting data that you need to invalidate because the flow is off.
One symptom of a leak in your system can appear when you perform a flow audit.  When the flow reported by the standard is more than ±2.5% from the flow that is being displayed by the instrument, you may have a leak.
Before attempting a leak check, you should first perform a flow calibration. If you are unable to calibrate the flow or your flow calibration constantly drifts, than this is a sign that you might have a leak.
To perform a leak check, you will need a NIST flow standard such as a BGI Delta Cal (Figure 1) or Streamline Pro.  You will also need  the flow audit adapter (Figure 2) sometimes referred to as a leak check adapter (PN.110524-00) that came with your instrument.
Don't miss out. Subscribe today.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.
Subscribe to Talk Tech ›
UX DATA-TRKR Retired - Admins, feel free to remove this reference component.