Meat species detection, screening & quantification
Imegen™ RapidFinder™ Meat ID Detection Kits have been developed in collaboration with Imegen to create a broad portfolio of real-time PCR-based animal species detection and quantification solutions. Among others, the range includes assay kits for beef, poultry, ruminant, equine and pork DNA in food and feed samples. The assays provide very high sensitivity with a limit of detection (LOD) below the officially required level of 0.1%.
Popular meat PCR testing products
RapidFinder™ Beef ID Kit
RapidFinder™ Poultry ID Kit
RapidFinder™ Pork ID Kit
These screening kits are designed for use with Applied Biosystems™ Nucleic Acid Extraction Kits, Applied Biosystems GMO Extraction Kit and Applied Biosystems 7500 Fast Real-Time PCR instrument. The RapidFinder™ Quant Multi-Meat Set, when used with one or more of the RapidFinder™ Meat ID kits, enables the simple and reliable determination of the percentage of a target meat species with respect to total meat in food and feed samples. The method involves detection of target species DNA by real-time PCR.
Resources
Follow us
Join our food testing community for the latest food safety news and updates