RapidFinder STEC Detection Workflow
Rapid, simple and accurate simultaneous detection of E. coli O157:H7 and the 'Big 6' non-O157:H7 STEC
The RapidFinder™ STEC Detection Workflow is a complete, validated screening and confirmation system for E. coli O157:H7 and the six non-O157 STEC (Shiga toxigenic E. coli).
Choose the best option for STEC detection
For beef producers, rapid and accurate results when screening products for STEC and other pathogens can bring substantial economic benefits.
  • Release product sooner
  • Avoid unnecessary product diversions and production shutdowns
  • Minimize risk to your brand and reputation
Current reference methods for detecting STEC in beef samples are accurate and sensitive but can be slow and labor-intensive. Alternative rapid methods can show reduced accuracy and often require large volumes of proprietary media, making them a risky and uneconomic option.
The Applied Biosystems™ RapidFinder™ STEC Detection Workflow enables screening for E. coli O157:H7 and six strains of Shiga toxigenic E. coli—the Big 6 STECs—in as few as 12 hours, providing a rapid, validated workflow for STEC detection in raw ground beef and raw beef trim.
The RapidFinder STEC Detection Workflow is a two-stage real-time PCR method, designed to accurately detect and confirm the presence of E. coli O157:H7 and the 'Big 6' serogroups of non-O157 STEC (serogroups O26, O45, O103, O111, O121 & O145), defined by the USDA Food Safety and Inspection Service (FSIS) as adulterants in the American beef industry.
Gain the benefits of the RapidFinder STEC Detection Workflow 
  • Greater confidence in results—optimized workflow, co-developed with USDA ARS, showed equivalent performance to the USDA-MLG reference method in AOAC Research Institute (AOAC-RI) validation studies
  • Simplified workflow—simultaneous detection of E. coli O157:H7 and any of the Big 6 non-O157:H7 E. coli in 375g raw ground beef and raw beef trim samples, with a system to fit within your existing testing program
  • Rapid product release—same-day detection of pathogenic STECs and clearance of STEC-negative product in as few as 12 hours with presumptive positives confirmed with just one additional PCR test (~1 hour)
  • Minimized costs, space requirements and handling steps—single enrichment step in non-proprietary enrichment broth
Two rapid, reliable alternative workflows to suit your food production
The RapidFinder STEC Detection System has two AOAC Research Institute (AOAC-RI) Performance Tested MethodsSM certified workflow options that greatly reduce the sample enrichment time and volume, adding a second layer of specificity for sero-group specific detection.
Workflow 1, ~12 hours, suitable for laboratories running a high test throughput
Automated sample preparation is used on the Applied Biosystems™ MagMax™ Express-96 Magnetic Particle Processor and Applied Biosystems™ PrepSEQ™ Nucleic Acid Extraction Kit. Up to 96 samples can be processed together, from enrichment to RapidFinder STEC Screening Assay results, in approximately 12 hours. One additional hour is required for the RapidFinder STEC Confirmation Assay results.  
Workflow 1, ~12 hours, suitable for laboratories running a high test throughput
Workflow 2, ~18 hours, ideal for lower test throughput 
The Applied Biosystems™ PrepSEQ™ Rapid Spin Sample Preparation Kit – Extra Clean with Proteinase K is used for manual PCR sample preparation with spin columns, providing screening release in approximately 18 hours.
Workflow 2, ~18 hours, ideal for lower test throughput
Learn more about the RapidFinder STEC Detection Workflow, and other products in our range for STEC testing, by reviewing the resources below or contact us to speak to one of our specialists.
Request more information
Food-Pathogen-PCR-Test-Selection-Guide-270x195
Choose the right PCR pathogen detection assays for your pathogen targets and sample types
Empower your operations with rapid reporting and detect foodborne pathogens quickly and accurately.  Our Food Pathogen PCR Test Selection Guide helps you to choose your validated workflow solution by pathogen target and sample type.
View selection guide
 Related Products Order code
Applied Biosystems™ PrepSEQ™ Nucleic Acid Extraction Kit
4428176
PrepSEQ Rapid Spin Sample Preparation Kit – Extra Clean with Proteinase K
4426715
PrepSEQ Nucleic Acid Extraction Kit*
4428176
PrepSEQ Nucleic Acid Extraction Kit*
4480466
RapidFinder STEC Screening Assay
4476886
RapidFinder STEC Confirmation Assay
4476901
RapidFinder STEC Screening Assay Starter Kit – with PrepSEQ Nucleic Acid Extraction Kit
4480467
7500 Fast Instrument Package, Laptop**
A30299
7500 Fast Instrument Package, Desktop**
A30304 
RapidFinder Express v2.0 and SDS 1.4.2 Software for 7500 Fast Instrument
A28811
*Used with the MagMax Express 96 Deep Well Processor
** Includes: 7500 Fast instrument, Computer, RapidFinder Express v2.0 and SDS 1.4.2 Software, capping tool, Precision plate holder, Calibration kits and Block balance tubes
Resources
Follow us
Join our food testing community for the latest food safety news and updates.