Perfect your food pathogen testing workflow
The Pathatrix® instrument is a fully automated sample prep system designed to rapidly and accurately handle any sample matrices to help save you time and money. Using paramagnetic particles that are either antibody-coated or positively charged, the Pathatrix system selectively binds and concentrates target organisms from many of the complex matrices found in food, environmental, and biothreat samples.
Request more info or a demo
Watch our free on-demand webinar | Request a Pathatrix demo in your lab | Pathatrix brochure  |  Pathatrix products
The Pathatrix Auto System is fully compatible with a wide range of detection technologies. The captured, concentrated, and cleaned sample produced by the Pathatrix system helps ensure a more reliable and accurate result from your chosen detection method. The time to result is also reduced due to the concentration effect of the Pathatrixsystem. Simple linking protocols are available for all of the commonly used detection technologies, including:
  • Agar plates—selective and chromogenic
  • Colorimetric assays—ELISA and lateral flow
  • PCR—real-time or conventional
Key features and benefits
Cleans and concentrates target—helps reduce false positives and indeterminate results
Pooling of up to 10 samples—save up to 90% on downstream detection costs
15-minute, fully automated run-time—quicker time to results and improved productivity
Resources
Pathatrix User Manual
Quick Reference Protocols
Pathatrix System Workflow
Pathatrix System—Generic Bead Beating Protocol
Pathatrix System—VIDAS® Strip Linking Step
Creating a Salmonella Culture Serial Dilution
Pathatrix system—BAX® Lysis Linking Step
Support
Phone and Email