Quantifiler® Duo DNA Quantification Kit
The Quantifiler® Duo DNA Quantification Kit enables forensic laboratories to simultaneously obtain a quantitative and qualitative assessment of total human and human male DNA in a single, highly sensitive real-time PCR reaction. This guides selection of the optimal STR chemistry (autosomal, Y-STR or miniSTR) and streamlines the workflow while increasing downstream analysis success rates.
Optimized to predict and improve performance with the AmpFℓSTR® PCR Amplification Kits, the Quantifiler® Duo kit integrates the forensic DNA workflow to maximize the recovery of interpretable STR profiles from sexual assault cases and other challenging samples.
Key Features
  • Highly Accurate: determines the ratio of human male to female DNA, even with minute male and excess female DNA (1:1000 or greater).
  • Highly Informative
  • Guides selection of the optimal AmpFLSTR® PCR Amplification Kit (autosomal, Y-STR or miniSTR)
  • Improves prediction of STR amplification performance and estimation of DNA quantity in degraded samples
Learn More
Product Categories
PRODUCT INFORMATION
Product Information
Integrated Workflow
Integrated Workflow
WHY USE Y?
Why Use Y?