View Important Notice >
Autosomal Locus Guide
AmpFℓSTR® kit locus reference table showing loci amplified by kit, dye label, and probability of identity.
Download a printable version.
Locus/
Chromosome Location
Identifiler®, Identifiler® Plus, & Identifiler® Direct Kits MiniFiler™ Kit NGM™ Kit NGM Select™ Kit Profiler Plus®, & Profiler Plus® ID Kits COfiler® Kit
SGM Plus® Kit
SEfiler Plus™ Kit Profiler® Kit
CSF1PO
5q33.3-34
6-FAM™ PET®       JOE™     JOE™
D5S818
5q21–31
PET®       NED™       NED™
D7S820
7q11.21–22
6-FAM™ 6-FAM™     NED™ NED™     NED™
D13S317
13q22–31
VIC® 6-FAM™     NED™       NED™
TPOX
2p23–2per
NED™         JOE™     JOE™
D3S1358
3p
VIC®   PET® PET® 5-FAM™ 5-FAM™ 5-FAM™ 6-FAM™ 5-FAM™
D8S1179
8q24.13
6-FAM™   VIC® VIC® JOE™   JOE™ VIC®  
D16S539
16q24–qter
VIC® NED™ 6-FAM™ 6-FAM™   5-FAM™ 5-FAM™ 6-FAM™  
D18S51
18q21.3
NED™ NED™ VIC® VIC® JOE™   JOE™ PET®  
D21S11
21q11.2–q21
6-FAM™ VIC® VIC® VIC® JOE™   JOE™ PET®  
FGA
4q28
PET® PET® NED™ NED™ 5-FAM™   NED™ NED™ 5-FAM™
TH01
11p15.5
VIC®   NED™ NED™   JOE™ NED™ NED™ JOE™
vWA
12p12–pter
NED™   6-FAM™ 6-FAM™ 5-FAM™   5-FAM™ 6-FAM™ 5-FAM™
D2S1338
2q35–37.1
VIC® VIC® 6-FAM™ 6-FAM™     5-FAM™ 6-FAM™  
D19S433
19q12–13.1
NED™   NED™ NED™     NED™ NED™  
D1S1656
1q42.2
    PET® PET®          
D2S441
2p14
    PET® PET®          
D10S1248
10q26.3
    6-FAM™ 6-FAM™          
D12S391
12p13.2
    PET® PET®          
D22S1045
22q12.3
    NED™ NED™          
SE33
6q14
      PET®       VIC®  
Amelogenin
X: p22.1–22.3
Y: p11.2
PET® VIC® VIC® VIC® JOE™ JOE™ JOE™ VIC® JOE™
Probability of Identity
African American* 1.31 x 10–18 6.52 x 10–11 6.52 x 10–20 1.75 x 10–21 1.48 x 10–11 3.15 x 10–7 7.91 x 10–14 6.47 x 10–15 1.23 x 10–10
US Caucasian* 5.01 x 10–18 8.21 x 10–11 2.78 x 10–19 4.07 x 10–21 1.04 x 10–11 1.19 x 10–6 2.99 x 10–13 7.46 x 10–14 2.79 x 10–10
US Hispanic* 7.65 x 10–18 1.05 x 10–10 2.59 x 10–19 6.62 x 10–21 NA NA NA NA NA
Native American* 3.62 x 10–17 2.08 x 10–10 NA NA NA NA NA NA NA
* Refer to appropriate User's Manual for additional population information.
† Based on preliminary calculations. Please refer to final User's Manual for complete information.
Y Chromosome Locus Guide
Locus Yfiler® Kit
DYS456 6-FAM™
DYS389I 6-FAM™
DYS390 6-FAM™
DYS389II 6-FAM™
Locus Yfiler® Kit
Y GATA H4 PET®
DYS437 PET®
DYS438 PET®
DYS448 PET®
Locus Yfiler® Kit
DYS458 VIC®
DYS19 VIC®
DYS385 a/b VIC®
Locus Yfiler® Kit
DYS393 NED™
DYS391 NED™
DYS439 NED™
DYS635 NED™
DYS392 NED™