Oil & Gas Testing
Proven analytical instruments
Petrochemical analysis can be challenging due to difficult matrices, a wide range of contaminants, and sensitivity requirements. We offer the broadest portfolio of laboratory analytical instrumentation in the industry that work together to enhance your analytical capabilities and streamline your processes.
Featured oil and gas testing products
Key processes
Product analysis of upper, middle, and downstream sectors include exploration, transportation and distillation.
Natural gas testing, including natural gas liquid, liquid petroleum gas, refinery gas, and specialty gas testing for hydrocarbons ranging from C1 to C6+ hydrocarbons.
Preventative maintenance analysis of scrubber solutions, boiler/cooling water and wastewater analysis. 
Industrial water processes keep the refinery operational. They include the use, treatment, and analysis of scrubber solutions and boiler/cooling water.
Digital Rock solutions provide insights that help operators and geoscientists improve reservoir characterization while also aiding cost-effective ultimate recovery and enhancing production predictability in a variety of conventional and unconventional plays.
Petroleum and natural gas workflows
Petroleum and natural gas workflows
Exploration and production
The upstream oil and gas sector encompasses both exploration and production of the fuels. Specifically, it includes searching for potential underground or underwater crude oil and natural gas fields, exploratory drilling, and established drill operations that bring the natural gas and crude oil to the surface.
Figure 1. The distribution of terpanes (biomarkers used in estimating the economics of crude by analyzing the oil-source rock as an indicator of source, maturity, migration, and biodegradation) in the aliphatic hydrocarbon fraction of crude oil samples. Analysis was performed using a Thermo Scientific™ DFS™ high resolution GC-MS (or equivalent Q Exactive GC Orbitrap GC-MS/MS) equipped with a multiple ion detector.
Figure 2. Determination of the boiling point range of gasoline with ethanol by ASTM D7096 simulated distillation. Samples were analyzed using a TRACE 1300 Series GC.
The upstream oil and gas sector refers to exploration and production. This  sector includes searching for potential underground or underwater crude oil and natural gas fields, exploratory drilling, and established drill operations that bring the natural gas and crude oil to the surface.
Recommended products
Transportation and custody transfer
The midstream oil and gas sector refers to the transportation, storage, and wholesale marketing of natural gas, crude, or refined petroleum products. Pipelines, rail cars, and ships are used to move crude oil and natural gas from production sites to refineries and deliver the various refined products to downstream distributors.
Any time natural gas changes hands - from initial release at the drill site until it reaches the end user - it is tested twice, by both the seller and buyer. The test results are compared as a critical quality control step in the process.
Figure 3. Determination of the calorific value of natural gas liquids (NGLs) by a TRACE 1300 Series GC with a thermal conductivity detector by GPA Method 2177. 
Figure 4. Analysis of corrosive oxygenates in LPGs and other light hydrocarbons samples by a TRACE 1310 Series GC by ASTM D7423. 
Recommended products
Refining, petrochemical, energy
The downstream oil and gas sector covers the refining of petroleum crude oil, the processing and purifying of raw natural gas, and distribution of petroleum or gas. Products are divided into light distillates (LPG C3-C4), gasoline (C3-C12, naphtha C6-14), middle distillates (kerosene, diesel C9-C16, jet fuel C9-C17) and heavy distillates (lubricating oil C20+).
Figure 5. Analysis of aromatics (used to boost octane levels in gasoline) is shown without backflush (left) and with backflush (right). In the right chromatogram the heavier matrix components are eliminated, leaving the column and detector cleaner. Analysis was performed using a TRACE 1310 Series GC with FID detector and Thermo Scientific™ TraceGOLD™ GC capillary column.
Figure 6. Analysis of trace elements in naphtha. Results for the spiked naphtha with relative standard deviation % (RSD %) of the three replicate measurements are within acceptable limits.
Figure 7. The separation of corrosive chlorine and sulfur in petroleum samples by CIC using a Thermo Scientific™ Dionex™ IonPac™ AS15 anion exchange column with suppressed conductivity detection. 
Recommended products
Resources
Application Notes
Case Studies
Brochures
Application Notebook
Articles