LC-MS offers versatility and resolution
LC-MS offers versatility and resolution
Liquid chromatography (LC) is a widely used method of sample ionization prior to analysis and is frequently coupled with mass spectrometry. With LC-MS, solubilized compounds (the mobile phase) are passed through a column packed with a stationary (solid) phase. This effectively separates the compounds based on their weight and affinity for the mobile and stationary phases of the column. This also leads to fragmentation of the sample and its anionization through loss of H+ ions.
Following this step, the sample passes into the vacuum chamber of the mass spectrometer.
LC is the separation technique of choice for larger and non-volatile molecules such as proteins and complex peptides. When combined with MS, LC-MS offers broad sample coverage because different column chemistries, such as reversed phase liquid chromatography, can be used.
LC is also an ideal method for separating isomers, which have the same mass and will otherwise not be differentiated (i.e., resolved) by a mass spectrometer. In fact, due to its superior resolving power and broad mass range, LC has largely replaced gel electrophoresis for molecular separation. Finally, LC helps reduce ion suppression, which occurs when molecules interact with one another and impede the process of complete ionization.
HPLC, which is defined as high performance liquid chromatography, has improved upon and largely replaced LC. HPLC was initially defined as high pressure liquid chromatography because it operates at a higher pressure ranging from 50-350 bar. In contrast, LC relies on gravity for the passage of the mobile phase through the column.
In this section, you will:
  • Learn which mass spectrometry instruments and technologies are best suited to sample analysis by LC-MS.
  • Understand how LC-MS helps analyze non-volatile and complex molecules.
  • Learn about workflows specifically designed for samples undergoing analysis by LC-MS.
LC-MS features
Technical handbook
Download our clinical research and forensic toxicology application handbook, which offers many LC-MS applications.
eBook
Understand recent UHPLC advances, the basics of glycan analysis, and intact antibody characterization.
eBook
Removing doubt — mass spectrometry workflows solving forensic cases.
eBook
Mass spectrometry workflows for improved healthcare.
LC-MS information subtopics
LC-MS Sample Types
Learn how LC-MS enables the analysis of complex, non-volatile and thermally labile samples including peptides, lipids, alcohols and steroids.
Mass Spectrometry Resource Library
Access our extensive collection of mass spectrometry workflows, scientific applications, technical handbooks, webinars and more. 