Your metabolic tools for drug discovery & development GIBCO® microsomes & other liver subcellular fractions
  • Large pooled lots for reproducible, long-term studies
  • Fractions carefully isolated by experts in hepatic biology
  • Microsomes functionally tested for select CYP450 and Phase II enzyme activities
Request more information
Human microsome pools and subcellular fractions
Subcellular fractions, derived from the endoplasmic reticulum of liver, contain a rich variety of metabolic enzymes for assessing the in vitro metabolism of drug candidates (Table 1) and are suitable for a variety of experiments:
  • Cytochrome P450 inhibition studies
  • Metabolic stability
  • Cytochrome P450 phenotyping
  • Metabolite characterization
50 donor human liver microsome pool
Designed to maximize functionality for all your study types- suitable for inhibition, clearance, or metabolic identification.
  • Equal representation of all 50 donors
  • Activities representing the typical patient population
11 specialty human liver microsome pools
Prepared with 5 to 20 donors each and pooled for your convenience according to age, gender, body mass index, high 3A activity, and high 2D6 activity.
Customized microsome pools
Our scientists will create a customized pool specifically for your unique research needs using our large, fully characterized single-donor database.
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1),  and trout (Oncorhynchus mykiss).  Custom species are available upon request. 
Each product contains an average representative pool of donors.  Human microsome pools are fully characterized (Km and Vmax) according to GLP standards for major cytochrome P450 activities and select Phase II enzymes using FDA recommended substrates (Tables 2-3).
Table 1. Relevant applications for liver subcellular fractions.
Metabolic  Enzymes Liver Microsomes Liver S9 Fractions Liver Cytosol
Aldehyde Oxidase
  X
X
Cytochromes P450 (CYP) X
X
Flavin Monooxygenases (FMO)
X
X
Glutathione Transferase (GST)   X
Monamine Oxidase (MAO)
  X
Sulfurotrnasferase (SULT)
  X
X
Uridine Glucuronide Transferase (UGT)
X
X
Table 2. Cytochrome P450 and Phase II enzymes tested in Gibco human microsome pools.
Enzyme Monitored Marker Substrate Incubation (min) Protein Concentration (mg/ml) Metabolite
CYP1A1/2
Phenacetin
30
0.1
Acetaminophen
CYP2A6
Coumarin
5
0.025
7-Hydroxycoumarin
CYP2B6
Bupropion
20
0.25
Hydroxybupropion
CYP2C8
Paclitaxel
10
0.075
6α-Hydroxypaclitaxel
CYP2C9
Tolbutamide
20
0.1
Hydroxytolubutamide
CYP2C19
(S) Mephenytoin
30
0.1
4'-Hydroxymephenytoin
CYP2D6
Dextromethorphan
15
0.2
Dextrorphan
CYP2E1
Chlorzoxazone
20
0.1
6-Hydroxychlorzoxazone
CYP3A4/5
Midazolam 4
0.025
1'-Hydroxymidazolam
CYP3A4/5
Testosterone 7
0.05
6β-Hydroxytestosterone
CYP4A11
Lauric Acid
15
0.2
12-Hydroxydecanoic Acid
FMO
Methyl p-Tolylsulfide 15
0.05
Methyl p-Tolylsulfide
UGT
7-Hydroxycoumarin
30
0.2
7-Hydroxycoumarin Glucuronide
Table 3. Kinetic parameters for the Gibco 50 donor human liver microsome pool.
Isoform Metabolite Km(μM) Vmax (nmol/min/mg)
CYP1A2 Acetaminophen 78 0.73
CYP2A6 7-Hydroxycoumarin 1.1 0.53
CYP2B6 Hydroxybupropion 64 0.29
CYP2C8 6α-Hydroxypaclitaxel 5.5 0.15
CYP2C9 Hydroxytolbutamide 220 0.19
CYP2C19 4'-Hydroxymephenytoin 34 0.031
CYP2D6 Dextrophan 3.2 0.13
CYP2E1 6-Hydroxychlorzoxazone 70 1.4
CYP3A4 6β-Hydroxytestosterone 19 4.0
CYP3A4 1'-Hydroxymidazolam 1.6 1.1
Support & Resources