Use this comparison chart to see which model is right for you
Prevent expensive plant decontamination and shutdown. Thermo Scientific ASM™ IV Automatic Scrap Monitoring Systems are configurable radiation monitors to prevent radioactive contamination from entering scrap metal and metal processing workflows. All ASM-IV radiation detectors are suited for portal monitoring applications that require the lowest possible alarm thresholds.
Contact a sales representative
Model Radiation detection module Number of detectors Vertical coverage Detector dwell System detector volume Use cases
ASM IV 3KE 2 2 48” (1219mm) 15” (381mm) 47 Liters
(2880 in2 )
• Scrap processing—material in/out
ASM IV 6KV 4 4 96” (2438mm) 15” (381mm) 94 Liters
(5760 in2 )
• Scrap processing—material in/out
ASM IV 6KE
2
4
51” (1295mm)
33” (838mm) 103 Liters
(6336 in2 )
• Scrap processing—material out
• Mill/Foundry—material in/out 
ASM IV 12KV
4
8
102” (2591mm)
33” (838mm)
208 Liters (12672 in2 )
• Scrap processing—material out 
• Mill/Foundry—material in/out
ASM IV 18KV 6 12 153” (3886mm) 33” (838mm) 311 Liters (19008 in2 ) • Scrap processing—material out 
• Mill/Foundry—material in/out
ASM IV 6KD 2 4 33”
(838mm)
48” (1219mm) 94 Liters
(5760 in2
• Scrap processing—material out
• Mill/Foundry—material in/out
ASM IV 12KD 4 8 69” (1753mm) 48” (1219mm) 189 Liters (11520 in2 ) • Scrap processing—material out
• Mill/Foundry—material in/out
ASM IV 18KD 6 12 105” (2667mm) 48” (1219mm) 283 Liters (17280 in2 • Scrap processing—material out 
• Mill/Foundry—material in/out
ASM IV 24KD 5 16 141” (3581mm) 48” (1219mm) 378 Liters (23040 in2 • Scrap processing—material out 
• Mill/Foundry—material in/out
All ASM-IV models feature a history banner of recent events:
  • Displays abbreviated results from the 10 most recent scan displays for rapid review. Tap the desired occupancy icon to display the detailed scan information.
  • Color-coded banner indicator for rapid scan results
  • Records and identifies time, date, and event ID number & customer name
Automatic Scrap Monitoring System