HB-GENESYSSelectionGuide-1180x250-6430
Developed for users, designed for results
From classroom teaching, to routine measurements, to discovery of the next breakthrough material, Thermo Scientific GENESYS spectrophotometers are designed to meet the needs of today's modern laboratory.
Click the UV-Vis and Vis tabs below to see our instrument comparisons. 
  • Start taking measurements immediately with the simple-to-use, high-resolution, color-display start menu
  • Exchange samples with ease with large, accessible sample compartment
  • Save your data fast any way you want it with multiple data-export options
Request a quote or consult
For teaching labs, research labs and industrial QC requiring robust construction, ease-of-use and software capability
For advanced teaching labs, R&D, research and life science applications requiring higher throughput or temperature control. For advanced teaching labs, R&D, research and life science applications requiring higher throughput or temperature control. For laboratories requiring double beam with a reference cell position.
Click on the instrument titles for more information and specifications.
GENESYS 50
UV-Vis Spectrophotometer
GENESYS 150
UV-Vis Spectrophotometer
BioMate 160
UV-Vis Spectrophotometer
GENESYS 180
UV-Vis Spectrophotometer
FEATURES:
Spectral range UV-Vis UV-Vis UV-Vis UV-Vis
Optics Dual Beam Dual Beam Dual Beam Double Beam
Display 7-inch touchscreen 7-inch touchscreen, tiltable 7-inch touchscreen, tiltable 7-inch touchscreen, tiltable
Supports test tubes
Removable and washable sample compartment
4 and 8-cell changer compatible* Single-cell only
Sipper, Peltier, fiber probe compatible
A-T-C
Scanning
Quant, fixed, live display, OD600
Kinetics
Thermo Scientific VISIONlite software compatible
Wi-Fi USB adaptor, available
Snap-on printer, available
Life Science methods - Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays
Sample compartment
  • Accessible from top, front or side
  • Accommodates up to 100 mm cuvettes
  • Available accessory for test-tubes up to 25 mm diameter and 150 mm height
Accommodates (optional):
  • 8-position cell changer
  • 4-position cell changer (long path cells)
  • Peltier thermostatted cell holder (20–60 °C)
  • Sipper accessory
  • Fiber optic probe coupler
Accommodates (optional):
  • 8-position cell changer
  • 4-position cell changer (long path cells)
  • Peltier thermostatted cell holder (20–60 °C)
  • Sipper accessory
  • Fiber optic probe coupler
  • 8-position cell changer (standard)
Also accommodates (optional):
  • 4-position cell changer (long path cells)
  • Peltier thermostatted cell holder (20–60 °C)
  • Sipper accessory
  • Fiber optic probe coupler
Warranty 2 years** 2 years** 2 years** 2 years**
  For teaching labs, research labs and industrial QC requiring robust construction, ease-of-use and software capability For teaching labs, research labs and industrial QC requiring robust construction, ease-of-use and software capability For advanced teaching labs, R&D, and research applications requiring higher throughput or temperature control.
Click on the instrument titles for more information and specifications.
GENESYS 30
Vis Spectrophotometer
GENESYS 40
Vis Spectrophotometer
GENESYS 140
Vis Spectrophotometer
FEATURES:
Spectral range Vis Vis Vis
Optics Single Beam Dual Beam Dual Beam
Display 5-inch color screen 7-inch touchscreen 7-inch touchscreen, tiltable
Supports test tubes
Removable and washable sample compartment
4 and 8-cell changer compatible* Single-cell only Single-cell only
Sipper, Peltier, fiber probe compatible
A-T-C
Scanning
Quant, fixed, live display, OD600
Kinetics
Thermo Scientific VISIONlite software compatible
Wi-Fi USB adaptor, available
Snap-on printer, available
Life Science methods - Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays
Sample compartment
  • Accessible from top, front or side
  • Accommodates up to 100 mm cuvettes
  • Available accessory for test-tubes up to 25 mm diameter and 150 mm height
  • Accessible from top, front or side
  • Accommodates up to 100 mm cuvettes
  • Available accessory for test-tubes up to 25 mm diameter and 150 mm height
Accommodates (optional):
  • 8-position cell changer
  • 4-position cell changer
  • Peltier thermostatted cell
  • Sipper accessory
Warranty 2 years** 2 years** 2 years**
For teaching labs, research labs and industrial QC requiring robust construction, ease-of-use and software capability
For advanced teaching labs, R&D, research and life science applications requiring higher throughput or temperature control. For advanced teaching labs, R&D, research and life science applications requiring higher throughput or temperature control. For laboratories requiring double beam with a reference cell position.
Click on the instrument titles for more information and specifications.
GENESYS 50
UV-Vis Spectrophotometer
GENESYS 150
UV-Vis Spectrophotometer
BioMate 160
UV-Vis Spectrophotometer
GENESYS 180
UV-Vis Spectrophotometer
FEATURES:
Spectral range UV-Vis UV-Vis UV-Vis UV-Vis
Optics Dual Beam Dual Beam Dual Beam Double Beam
Display 7-inch touchscreen 7-inch touchscreen, tiltable 7-inch touchscreen, tiltable 7-inch touchscreen, tiltable
Supports test tubes
Removable and washable sample compartment
4 and 8-cell changer compatible* Single-cell only
Sipper, Peltier, fiber probe compatible
A-T-C
Scanning
Quant, fixed, live display, OD600
Kinetics
Thermo Scientific VISIONlite software compatible
Wi-Fi USB adaptor, available
Snap-on printer, available
Life Science methods - Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays
Sample compartment
  • Accessible from top, front or side
  • Accommodates up to 100 mm cuvettes
  • Available accessory for test-tubes up to 25 mm diameter and 150 mm height
Accommodates (optional):
  • 8-position cell changer
  • 4-position cell changer (long path cells)
  • Peltier thermostatted cell holder (20–60 °C)
  • Sipper accessory
  • Fiber optic probe coupler
Accommodates (optional):
  • 8-position cell changer
  • 4-position cell changer (long path cells)
  • Peltier thermostatted cell holder (20–60 °C)
  • Sipper accessory
  • Fiber optic probe coupler
  • 8-position cell changer (standard)
Also accommodates (optional):
  • 4-position cell changer (long path cells)
  • Peltier thermostatted cell holder (20–60 °C)
  • Sipper accessory
  • Fiber optic probe coupler
Warranty 2 years** 2 years** 2 years** 2 years**
  For teaching labs, research labs and industrial QC requiring robust construction, ease-of-use and software capability For teaching labs, research labs and industrial QC requiring robust construction, ease-of-use and software capability For advanced teaching labs, R&D, and research applications requiring higher throughput or temperature control.
Click on the instrument titles for more information and specifications.
GENESYS 30
Vis Spectrophotometer
GENESYS 40
Vis Spectrophotometer
GENESYS 140
Vis Spectrophotometer
FEATURES:
Spectral range Vis Vis Vis
Optics Single Beam Dual Beam Dual Beam
Display 5-inch color screen 7-inch touchscreen 7-inch touchscreen, tiltable
Supports test tubes
Removable and washable sample compartment
4 and 8-cell changer compatible* Single-cell only Single-cell only
Sipper, Peltier, fiber probe compatible
A-T-C
Scanning
Quant, fixed, live display, OD600
Kinetics
Thermo Scientific VISIONlite software compatible
Wi-Fi USB adaptor, available
Snap-on printer, available
Life Science methods - Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays
Sample compartment
  • Accessible from top, front or side
  • Accommodates up to 100 mm cuvettes
  • Available accessory for test-tubes up to 25 mm diameter and 150 mm height
  • Accessible from top, front or side
  • Accommodates up to 100 mm cuvettes
  • Available accessory for test-tubes up to 25 mm diameter and 150 mm height
Accommodates (optional):
  • 8-position cell changer
  • 4-position cell changer
  • Peltier thermostatted cell
  • Sipper accessory
Warranty 2 years** 2 years** 2 years**
** Three-year warranty available upon instrument registration
Legendary reliability. Unrivaled usability.
Built on the foundation of 60 years of spectroscopy experience, the next generation Thermo Scientific GENESYS family of Vis / UV-Vis spectrophotometers provides exceptional performance with a contemporary design. There is a GENESYS spectrophotometer you can count on to provide accurate, reproducible results for your application requirements. 
Download the brochure
GENESYS Family Brochure