Transmission
How does it work?
The transmission technique does not require a separate accessory. The user simply places a sample directly into the infrared (IR) beam. As the IR beam passes through the sample, the transmitted energy is measured and a spectrum is generated. However, the analyst must often prepare the sample into a pellet, mull, film, etc. before the transmission measurement can be made. This requires expertise and can be time consuming.
Watch the video series
What types of samples can you analyze?
Excellent quality spectra can be obtained for many types of samples using transmission. The transmission technique can be used alone or in conjunction with accessories such as microscopes and liquid or gas cells to analyze:
  • Organic powders in pellet or mull form
  • Thermoplastic powders
  • Soluble polymers
  • Thin polymer films
  • Regular-shaped polymers (with preparation)
  • Irregular-shaped polymers (with preparation)
  • Dark polymer films (not carbon-filled)
  • Liquids (free-flowing or viscous)
  • Gases (high concentrations to trace amounts)
What are the advantages of transmission?
  • Economical – cells and mounts are generally inexpensive
  • Well established – most traditional form of sample measurement
  • Excellent spectral information – ideal for qualitative measurements
  • Great for quantitative work – many standard operating procedures are based on transmission
Transmission - watch and learn with this video series
Basics
Sample preparation
Data collection
Sample preparation
Data collection
Sample preparation
Data collection
Introducing Mike
Dr. Michael Bradley received his BS degree in chemistry from the University of South Carolina and his PhD in physical chemistry from the University of Illinois, and also completed his MBA in management. He taught graduate and undergraduate chemistry for 15 years, prior to becoming a field applications scientist with Thermo Nicolet, subsequently Thermo Fisher Scientific, in 2002. He is now the marketing manager for FTIR and FTIR microscopy products.
Spectroscopy, Elemental & Isotope Analysis Resource Library
Access a targeted collection of application notes, case studies, videos, webinars and white papers covering a range of applications for Fourier Transform infrared spectroscopy, Near-infrared spectroscopy, Raman spectroscopy, Nuclear Magnetic Resonance, Ultraviolet-Visible (UV-Vis) spectrophotometry, X-Ray Fluorescence, and more.
Resources
Support
  • Knowledge Base
    Thermo Scientific Knowledge Base for Molecular Spectroscopy offers a web-based, self-help option for answering product questions, accessing documentation and resolving issues.
Share