AA Instruments
Be more efficient with more automation
High performance at great value and low cost of ownership. Perform efficient, accurate trace elemental analysis with these high-performance double beam, graphite furnace and dual-atomizer double beam AA spectrometers.
AA spectrometer selection guide
iCE 3300 AA
iCE 3300 GF AA
iCE 3400 AA
iCE 3500 AA
Atomizer Flame Furnace Furnace Flame & Furnace
Vapor atomization Optional
Furnace vision system No Optional Optional As standard
Carousel 6-lamp auto-aligning
Optics Single-beam Single-beam Double-beam, Stockdale & self-referencing Zeeman Double-beam, Stockdale & self-referencing Zeeman
Photomultiplier Standard & optional wide-ranging Standard & optional wide-ranging Wide-ranging
Wavelength range (nm) 185 - 760 185 - 760 180 - 900
Size (mm) W 575, H 527, D 595 W 575, H 527, D 595 W 788, H 527, D 595 W 788, H 527, D 595
Resources