Plant DNA Isolation kits
Plant DNA Isolation has unique challenges that require kits specifically designed to deal with carbohydrates, phenolics, and other compounds abundant in plant tissues. Plant cell walls can be very difficult to disrupt, and lysates often contain significant amounts of compounds such as tannins, phenolics, and complex polysaccharides that can affect DNA and RNA quality and inhibit downstream reactions. Life Technologies offers an expansive portfolio of kits, reagents, and devices for the isolation, purification, analysis, and quantitation not only for plant genomic DNA, but also for total RNA, mRNA, and microRNA or transcriptome RNA.
Jump to plant RNA Isolation
Which Plant DNA Isolation Kit is Right for You?
DNA extraction and TaqMan® chemistry-based genotyping combined, <1h to results
Magnetic beads designed to handle low-abundance plant DNA samples
Most suitable for screening and GMO testing due to automation capabilities
Slica-based method for low abundance plant DNA samples
Bulk buffers for crude extraction of DNA and RNA from plant cells (*)
The most efficient lysis for the largest amount of tissue
TaqMan® Sample-to-SNP™ Kit
MagMAX™ DNA Multi-Sample Kit
ChargeSwitch® gDNA Plant
PureLink™ Genomic Plant
Total RNA Lysis Solution
Plant DNAzol®
Starting Material
2-3mm punch
5-10 mg
50-100 mg
100 mg
≥ 100 mg
≥ 100 mg
Isolation technology
Lysis solution
Magnetic beads
ChargeSwitch® Chemistry, magnetic beads
Silica spin column
Bulk lysis buffer
Organic extraction
Downstream applications
Real-time PCR
All
All
All
PCR, real-time PCR
All
Automatable
yes
yes
yes
no
no
no
Yield
varies based on starting material
up to 14.6 µg
up to 7 µg
up to 14.6 µg
varies based on starting material
varies based on starting material
Protocol time
5 mins
<40 mins
<15 mins
<40 mins
<15 mins
<60 mins
Plant sample types
Works with broad rangeof plant tissues and starting material (leaf punch, needle, seed chips)
Chloroplasts and broad range of plant tissues (alfalfa and sunflower sprouts, corn husks, soybeans, mushroom, tomato leaves, wheat grass &Arabidopsis thalianaleaves)
Some of the many species and tissues are canola, corn, cotton, sunflower, barley, rice (seeds); tomato, cucumber, lettuce, banana (leaves); rose, magnolia (flower); Acer, sycamore, grass, etc.
Chloroplasts and broad range of plant tissues (alfalfa and sunflower sprouts, corn husks, soybeans, mushroom, tomato leaves, wheat grass andArabidopsis thalianaleaves)
Customer-validated for DNA on a broad range of plant tissues for SNP genotyping as well as corn flour for GMO testing
Broad range of plant tissues (alfalfa and sunflower sprouts, corn husks, soybeans, mushroom, tomato leaves, wheat grass andArabidopsis thalianaleaves)
High throughput compatibile
yes
yes
yes
no
yes
yes
High throughput/ 
automation options
yes
50 preps 
96 preps
ChargeSwitch® gDNA Plant 960 Kit
96-well plate
yes
yes
Kit sizes available
100 preps 
400 preps 
4000 preps
50 preps 
96 preps
96 preps 
960 preps
50 preps 
4 X 96 preps
250 mL
100 mL
Order now
Order now
Order now
Order now
Order now
Order now
(*)The Total RNA Lysis solution was part of the discontinued ABI PRISM® 6100 Nucleic Acid PrepStation and is our cheapest and quickest methodology for plant DNA extraction without the use of organic solvents.
Which Plant RNA Isolation Kit is Right for You?
  Great for difficult samples (conifer tissue & seeds) Rapid & fully automated Quick & easy to use Efficient recovery of microRNA & small RNA
   Plant RNA Reagent  MagMAX™-96 Total RNA Isolation Kit  PureLink® RNA Mini Kit  mirVana™ miRNA Isolation Kit
RNA types isolated large RNA molecules only large RNA molecules only large RNA molecules only Small & large RNA molecules
Prep time 60 minutes <45 minutes <20 minutes 30 minutes
Amount of starting material Up to 1 g Up to 10 mg <250 mg 0.5 to 250 mg
Isolation method High purity organic extraction
(requires alcohol precipitation)
Scalable, flexible format with magnetic beads Fast, convenient silica column Highest purity & convenience; includes organic extraction & silica column
(no precipitation required)
High throughput compatible   yes    
Compatible products   Ambion® Plant RNA Isolation Aid
Removes common plant contaminants
Ambion® Plant RNA Isolation Aid
Removes common plant contaminants
Ambion® Plant RNA Isolation Aid
Removes common plant contaminants
  Order Now Order Now Order Now Order Now
Related products
In addition to DNA and RNA isolation products, Thermo Fisher offers an expansive portfolio of solutions for analysis and quantitation of plant genomic DNA.