Due to its improved brightness, superior photostability, and extremely rapid maturation rate, the mCherry monomeric red fluorescent protein is becoming the red fluorescent protein of choice for monitoring physiological processes and detecting transgenic expression. This affinity-purified rat monoclonal antibody can be used to detect native and denatured forms of mCherry or mCherry fusion proteins in western blot analysis, immunoprecipitation, immunocytochemistry, and flow cytometry applications.
See all anti-mCherry antibodies
Validated for a variety of applications
  • Western blotting
  • Immunocytochemistry
  • Immunoprecipitation
  • Flow cytometry
  • Immunohistochemistry
In western blot analysis, this mCherry rat monoclonal antibody demonstrates no cross reactivity with Emerald GFP, TagGFP, mKate2, or TagRFP. The same degree of specificity is also demonstrated in immunocytochemistry and flow cytometry applications. Additionally, this antibody has been used successfully in immunoprecpitation.
IHC using an anti-mCherry antibody (fluorescent and stained image). Intestinal tissue from a transgenic mouse expressing mCherry in all tissues. Panel A) Fluorescent image detecting mCherry expression and Panel B) HRP-stained image. Following isolation and fixation in 4% paraformaldehyde, mCherry rat monoclonal antibody (Cat. No. M11217) was used at a 1:15,000 dilution. The ImmPRESS™ Anti-Rat Ig (peroxidase) Polymer Detection Kit (Vector Laboratories) was used according to manufacturer’s instructions; sections were incubated in peroxidase substrate solution until the desired stain intensity developed.
mCherry rat monoclonal antibody conjugates
Fluorescent conjugates of our affinity-purified rat monoclonal antibodies for mCherry fluorescent protein can be used to detect native and denatured forms of mCherry or mCherry fusion proteins in flow cytometry applications. Full-length mCherry was used as the immunogen, and the resulting monoclonal IgG2a-isotype antibody detects denatured and native forms of the protein. Because of its improved brightness, superior photostability, and extremely rapid maturation rate, mCherry monomeric red fluorescent protein is becoming the red fluorescent protein of choice for monitoring physiological processes and detecting transgene expression.
Available conjugates of mCherry rat monoclonal antibody (clone 16D7)
Dye conjugate Excitation laser color Excitation/emission (nm) Cat. No.
Pacific Blue dye Violet 405/455 M11238
Alexa Fluor 488 Blue 488/519 M11239
Alexa Fluor 594 Red 590/617 M11240
Alexa Fluor 647 Red 650/668 M11241
Histogram of gated U2-OS cells expressing mCherry protein and labeled with Pacific Blue dye-conjugated rat anti-mCherry antibody (Cat. No. M11238). Samples were acquired and analyzed using 405 nm excitation and 522/31 nm band pass emission filters on an Attune Flow Cytometer.
Resources
Product selection guides
Learning and support
Cell analysis resources