The HA (hemagglutinin) tag is derived from the human influenza virus HA protein. It has been extensively used as a general antibody epitope tag and is well-characterized. HA tag antibodies provide a dependable method for the detection and purification of tagged target proteins without a protein-specific antibody or probe.
Invitrogen HA tag antibodies reliably detect recombinant HA tagged proteins, and each antibody is validated for use in a variety of applications.
See all anti-HA tag antibodies
Featured product data
Western blot analysis of HA epitope tag. Analysis was performed by running various amounts of E. coli lysate containing a multi-epitope tagged protein per well in a 4-20% Tris-HCl polyacrylamide gel. Proteins were transferred to a nitrocellulose membrane and blocked with 5% BSA/TBST for at least 1 hour. The membrane was probed with an HA epitope tag monoclonal antibody (Cat. No. 26183) at a dilution of 1:10,000 for 1 hour at room temperature on a rocking platform, washed in TBS-0.1%Tween™ 20, and probed with a goat anti-mouse IgG-HRP secondary antibody (Cat. No. 32430) at a dilution of 1:20,000 for at least 1 hour. Chemiluminescent detection was performed using Thermo Scientific SuperSignal West Pico Chemiluminescent Substrate (Cat. No. 34080).
Immunofluorescent analysis of HeLa cells transfected with a construct containing an HA epitope tag. Formalin fixed cells were permeabilized with 0.1% Triton™ X-100 in TBS for 10 minutes at room temperature and blocked with 1% Thermo Scientific Blocker BSA (Cat. No. 37525) for 15 minutes at room temperature. Cells were probed with a DyLight 488-conjugated HA epitope tag monoclonal antibody (Cat. No. 26183-D488) at a dilution of 1:25 for at least 1 hour at room temperature. Nuclei (blue) were stained with Hoechst™ 33342 dye (Cat. No. 62249). Images were taken on a Thermo Scientific ArrayScan imager at 20x magnification.
Annotated product references
  • Cat. No. 26183 was used in immunoprecipitation to identify novel protein co-factors of the beta2 isoform of the thyroid hormone receptor. Molecular endocrinology (Baltimore, Md.) (May 2013; 27: 840) "Research resource: identification of novel coregulators specific for thyroid hormone receptor-β2." Hahm JB,Privalsky ML
  • Cat. No. 26183 was used in immunoprecipitation to evaluate a recombinant myeloperoxidase antibody. Analytical biochemistry (Mar 2011; 410: 1) "A high-affinity recombinant antibody permits rapid and sensitive direct detection of myeloperoxidase." McDonnell B,Hearty S,Finlay WJ,O'Kennedy R
  • Cat. No. PA1-985 was used in western blot to study the roles of PAN1 and PAN2 in the polarization of maize cell division. The Plant cell ( 2012; 24: 4577) "Identification of PAN2 by quantitative proteomics as a leucine-rich repeat-receptor-like kinase acting upstream of PAN1 to polarize cell division in maize." Zhang X,Facette M,Humphries JA,Shen Z,Park Y,Sutimantanapi D,Sylvester AW,Briggs SP,Smith LG
Resources
Product selection guides
Learning and support
Cell analysis resources