A container solution for every scale
Whether your application involves large volume pelleting, protein purification, DNA isolation, or other separation methods, our comprehensive selection of Nalgene™ centrifuge bottles offers a solution for nearly every scale. Our bottles are available in sizes ranging from 175 mL to 2 L, in a variety of materials, with round or conical bottom configurations.
Popular process centrifugation bottles
Nalgene™ 2L PPCO Bio Bottle with Sealing Closure
Nalgene™ 1L Super-Speed Polycarbonate Centrifuge Bottles with Sealing Closure
Nalgene™ Polycarbonate Centrifuge Bottles
Low Speed Centrifuge Bottles
Reliable, accurate labware. Economical and disposable solutions for low speed, large volume cell harvesting, pelleting and protein purification.
High Speed Centrifuge Bottles
Durable and guaranteed leakproof. Boost productivity and pellet recovery in high-speed, large-volume applications with leakproof, durable centrifuge bottles.
Centrifuge Bottle Accessories
High-quality and reliable. Maximize performance and ensure proper rotor fit of your centrifuge bottles with Nalgene centrifuge bottle sealing cap assemblies and Nalgene centrifuge bottle adapters.
Quality, choice, and innovation for your applications
Enhance productivity through innovative general labware products. Our proven lab consumables will meet your specialized application needs, and help you maintain a clean, safe, and efficient laboratory.
Resources