If you agree your CHO-derived biologic shouldn’t be subject to development constraints imposed by others, break free with Freedom® Cell Line Development Kits. Commercial licensing that’s simple, flexible and without royalties.
ProBioGen's logo
  • Commercial licensing that’s simple, flexible and without royalties (see figure 1) - Licensing FAQ's
  • Consistently generate IgG producing scalable clones that meet current industry standards for productivity (see figure 2)
  • Easy-to-use kit that can take your gene of interest from transfection to lead clone in typically less than 6 months
  • Completely integrated animal origin-free workflow utilizing a fully-documented cGMP parental cell line
  • Gibco® Freedom® CHO-S® Kit developed in collaboration with ProBioGen AG
Two kits are available, each backed by the technical support of Life Technologies’ Process Science Team
  • Not sure which kit best fits your needs?
  • Do you need help using your Freedom Cell Line Development Kit?
Contact the Life Technologies' Process Scientist Team
  • Do you want to learn more about our process development team?
Gibco® Freedom® CHO-S® Kit
Gibco® Freedom® DG44 Kit
Ability to achieve titers >3 g/L Proven host cell used in the production of commercial products
Proven in IgG production Can be used for IgG production
Attain scalable clones in <6 months Attain scalable clones in <6 months
Developed in collaboration with ProBioGen AG Proven in recombinant protein production
Learn more about ProBioGen AG Developed by Life Technologies
Gibco® Freedom® CHO-S® Kit FAQ's Gibco® Freedom® DG44 Kit FAQ's
View the Product View the Product
Not sure which kit is best for you? Ask our Process Scientists Team.
Licensing questions? Visit our Licensing FAQ's page or contact outlicensing@lifetech.com for license terms.
Figure 1: Freedom® Cell Line Development Kits Flexible Business Model   Figure 2: Industry-standard titers achieved with Freedom™ CHO-S® Kit
Freedom from royalties? √  Yes
Freedom from annual maintenance fees? √  Yes
Freedom to use any contract manufacturer √  Yes
Freedom to host cell line for multiple projects? √  Yes
Freedom to modify the host cell line? √  Yes
Freedom the modify the purchased vector? √  Yes
Freedom to conduct process optimization? √  Yes
Freedom to request support when desired? √  Yes
Freedom to partner with others within field of use?* √  Yes
Clone productivity was assessed using a DASGIP parallel bioreactor system in CD FortiCHO™ Medium supplemented with L-glutamine and CD EfficientFeed™ B. Cultures were fed with CD EfficientFeed™ C and glucose. Final clone productivity ranged between 1.5 and 3.3 g/L.
Maximize Your Gibco® Freedom® CHO-S™ Kit Experience
For all users of the Gibco® Freedom® CHO-S™ Kit, we’ve developed a special FAQ eBook to enable you to get the most success from your kit. Inside, you’ll find the most commonly asked questions answered directly by our kit experts.
The FAQ eBook has been specially formatted to provide an incredible, interactive experience.  Each FAQ section contains:
  • Frequently asked questions (FAQs)
  • Dos and Don'ts
  • Helpful tips
  • Illustrative photos, diagrams and video
Download your copy of the Gibco® Freedom® CHO-S™ Kit FAQ eBook
Gibco® Freedom® CHO-S™ FAQ
Please note that the content in the FAQ document is not meant to replace the full User Guide. To download the full User Guide, please click the link below:
Gibco® Freedom® CHO-S™ Kit users guide