Research Scale Purification graphic
The research portfolio comprises of a unique set of affinity resins for purification of a variety of antibody formats and species, a number of resins for purification of biosimilars, an innovative tag system based on only 4 amino acids and a set of products for plasma sample preparation.
Request more information ›       Order Now
  • Antibody Purification
    Purify virtually any antibody format from any species using our diverse product portfolio
  • C-tag
    Smallest affinity tag system (4 amino acid recognition) available on the market, offering high selectivity, affinity, and a variety of elution conditions
  • POROS™ metal chelate columns
    Prepacked Immobilized Metal Affinity Column is based on an imidodiacetate functional group designed for the purifiction of proteins that form co-ordination complexes via metal sites and specific amino acids, such as histidine tagged fusion proteins.
  • Plasma sample preparation
    Single-step, single-use depletion of highly abundant proteins from human serum and plasma, 99% protein removal before 2D gel or Mass spec analysis.
  • POROS™ HQ 10 & 20 µm Columns
    Prepacked Strong Anion Exchange Column is based on a quarternized polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography™ media designed for the separation and purification of biomolecules.
  • POROS™ PI 20 µm Columns
    Prepacked Weak Anion Exchange Column is based on a polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography™ media designed for the separation and purification of biomolecules.
  • POROS™ HS 20 µm Columns
    Prepacked Strong Cation Exchange Column is a high capacity, strong cation exchange Perfusion Chromatography™ media suitable for the separation and purification of biomolecules.
  • POROS™ S 10 & 20 µm Columns
    Prepacked Strong Cation Exchange Column is a strong cation exchange Perfusion Chromatography™ media, suitable for the separation and purification of biomolecules.
  • POROS™ R2 10 & 20 µm Columns
    Pre-packed Reversed-Phase Column is a 2000 angstrom pore size poly (styrene-divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS™ R1 10 & 20 µm Columns
    Pre-packed Reversed-Phase Column is a 4000 angstrom pore size Poly (Styrene-Divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS™ HP2 20 µm Columns
    Prepacked Hydrophobic Interaction Chromatography Column is a high density phenyl, Perfusion Chromatography™ media suitable for the separation and purification of proteins.
  • Antibody Purification
    Purify virtually any antibody format from any species using our diverse product portfolio
  • C-tag
    Smallest affinity tag system (4 amino acid recognition) available on the market, offering high selectivity, affinity, and a variety of elution conditions
  • POROS™ metal chelate columns
    Prepacked Immobilized Metal Affinity Column is based on an imidodiacetate functional group designed for the purifiction of proteins that form co-ordination complexes via metal sites and specific amino acids, such as histidine tagged fusion proteins.
  • Plasma sample preparation
    Single-step, single-use depletion of highly abundant proteins from human serum and plasma, 99% protein removal before 2D gel or Mass spec analysis.
  • POROS™ HQ 10 & 20 µm Columns
    Prepacked Strong Anion Exchange Column is based on a quarternized polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography™ media designed for the separation and purification of biomolecules.
  • POROS™ PI 20 µm Columns
    Prepacked Weak Anion Exchange Column is based on a polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography™ media designed for the separation and purification of biomolecules.
  • POROS™ HS 20 µm Columns
    Prepacked Strong Cation Exchange Column is a high capacity, strong cation exchange Perfusion Chromatography™ media suitable for the separation and purification of biomolecules.
  • POROS™ S 10 & 20 µm Columns
    Prepacked Strong Cation Exchange Column is a strong cation exchange Perfusion Chromatography™ media, suitable for the separation and purification of biomolecules.
  • POROS™ R2 10 & 20 µm Columns
    Pre-packed Reversed-Phase Column is a 2000 angstrom pore size poly (styrene-divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS™ R1 10 & 20 µm Columns
    Pre-packed Reversed-Phase Column is a 4000 angstrom pore size Poly (Styrene-Divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS™ HP2 20 µm Columns
    Prepacked Hydrophobic Interaction Chromatography Column is a high density phenyl, Perfusion Chromatography™ media suitable for the separation and purification of proteins.
Related products and services
Application Notes and Webinars
Bioprocessing resources
Click below to access products brochures, training videos, mobile apps, and more.
Support
 Request more info on Bioprocessing