Three60 Single-Use Sampling System
Rapid, safe, sterile sampling system
Single-use systems enable improved sterility and reduced cleaning requirements over traditional sampling systems across all applications in the biopharmaceutical industry. Compared to traditional sampling systems, the Thermo Scientific Three60 Single-Use Sampling System brings additional advantages to the sampling process, resulting in a quicker solution and a safer process.
Request information Download product brochure
Design features of the Three60 sampling system
The simple design behind the Three60 Sampling System allows you to take a representative sample of your product with minimal effort. For a completely sterile transfer, use a clean-in-place (CIP) or sterilization-in place (SIP) process to prepare your tank. The pre-irradiated BioProcess Containers (BPCs) and assemblies will ensure a sterile fluid path, while the quick-turn valve and pinch-and-cut disconnections maintain sterile transfer and removal.
Key features:
  • No parts to be assembled, disassembled, and cleaned
  • No additional tools or expensive hardware required
  • Each package contains one valve and four assemblies with pinch-and-cut disconnections
  • The sanitary connector 3.8 cm (1.5 in.) is compatible with all vessels
  • Up to four samples can be removed within minutes
Valve mechanics
The Three60 valve only turns in one direction and will only turn once to an open position then again to a closed position for each line. There is no risk of cross-contamination between lines because of the single-turn direction of the valve barrel and the individual fluid paths for each sample assembly. Additionally, the valve closes ensuring that no fluid “hold-up” can be flushed into the next sample during the CIP/SIP process.
From kit to tank:
  • Quick-turn Three60 valve: The face of the Three60 valve can be sterilized with the tank through traditional CIP/SIP processes. The valve has four sterile assemblies. This helps maintain equipment, product, and sample sterility throughout the process.
  • Pinch-and-cut sterile disconnections: The pinch-and-cut disconnections allow the technician to quickly collect the sample and eliminate the need for additional tools or tube sealing.
  • Injection ports: The luer lock injection site permits extraction through either the septum or twist-off luer fitting to pour.
  • Pre-irradiated BPC assemblies: The Three60 system is provided with pre-irradiated BPC assemblies in sizes ranging from 50 mL to 2 L.
The simple design behind the Three60 Sampling System allows you to take a representative sample of your product with minimal effort. For a completely sterile transfer, use a clean-in-place (CIP) or sterilization-in place (SIP) process to prepare your tank. The pre-irradiated BioProcess Containers (BPCs) and assemblies will ensure a sterile fluid path, while the quick-turn valve and pinch-and-cut disconnections maintain sterile transfer and removal.
Key features:
  • No parts to be assembled, disassembled, and cleaned
  • No additional tools or expensive hardware required
  • Each package contains one valve and four assemblies with pinch-and-cut disconnections
  • The sanitary connector 3.8 cm (1.5 in.) is compatible with all vessels
  • Up to four samples can be removed within minutes
Valve mechanics
The Three60 valve only turns in one direction and will only turn once to an open position then again to a closed position for each line. There is no risk of cross-contamination between lines because of the single-turn direction of the valve barrel and the individual fluid paths for each sample assembly. Additionally, the valve closes ensuring that no fluid “hold-up” can be flushed into the next sample during the CIP/SIP process.
From kit to tank:
  • Quick-turn Three60 valve: The face of the Three60 valve can be sterilized with the tank through traditional CIP/SIP processes. The valve has four sterile assemblies. This helps maintain equipment, product, and sample sterility throughout the process.
  • Pinch-and-cut sterile disconnections: The pinch-and-cut disconnections allow the technician to quickly collect the sample and eliminate the need for additional tools or tube sealing.
  • Injection ports: The luer lock injection site permits extraction through either the septum or twist-off luer fitting to pour.
  • Pre-irradiated BPC assemblies: The Three60 system is provided with pre-irradiated BPC assemblies in sizes ranging from 50 mL to 2 L.
Related products
Standard and custom BPCs, including our three main film platforms (Aegis5-14, CX5-14, and ASI 26/77), tubing sets, and manifolds, covering a range of applications, sizes, and connection systems.
Bioreactor platforms designed as stirred-tank reactors for animal cell culture, utilizing single-use product contact surfaces.
Ordering information
Resources
Three60 Sampling System
Bioprocessing resources
Click below to access products brochures, training videos, mobile apps, and more.
Support
 Request more info on Bioprocessing