carboys_230x195px
The Thermo Scientific Nalgene carboy advantage
When using carboys for critical applications, standard laboratory carboys won't do. More supporting documentation, choices and designs sensitive to the necessary cleaning validations are just a few things to look for. Customization with fluid transfer options, pre-sterilization, and forced extraction studies are additional benefits you can expect from Thermo Scientific Nalgene carboys and jerricans. 
Request information
Key advantages of the Nalgene Carboys
  • Well supported with off-the-shelf, validation binders, and extractables studies 
  • Screw top, leakproof seals to ensure sample integrity with sanitary fitting options available for easier cleaning validations
  • Available in LDPE, HDPE, Fluorinated, PP, PC, up to 50 L
  • Rectangular options for dense storage efficiency and round options to facilitate stirring
  • Ability to provide custom tubulation for specific fluid transfer requirements
  • Sterile, single-use option can eliminate the need for in-house packaging and sterilization 
Find your production carboy for your application by resin type
LDPE Carboys
Suited for advanced liquid handling and solution storage for many pharmaceutical and biotechnology applications, these single-use carboys are the ideal choice for storage and transport of reagents. These containers can eliminate cleaning costs, validation and assembly, and contamination from repeated use.    
HDPE Carboys and Jerricans
Manufactured for tough applications, these carboys and jerricans are suited for storing and dispensing solutions and media. We offer a selection of heavy-duty, fluorinated and HDPE jerricans graduated in liters and gallons. 
These Nalgene labware are sterilized and tested after assembly, with sterility guaranteed as long as the packaging is intact. Indicator dots used on Nalgene products undergo an irreversible color change upon sterilization.    
Polypropylene Carboys
Durable, lighter and safer than glass, these carboys are available in a variety of shapes and sizes, with a range of features such as wide mouths and handles for convenient filling and handling
Easier to clean than threaded containers or vessels, these one-piece molded, non-threaded carboys are designed for use as receiver or dispensing vessels in biopharmaceutical applications. Solid ported 
Polycarbonate Carboys
Our polycarbonate carboys ensure the integrity of your samples. Glass-clear to permit easy visual inspection and volume measurements but lighter and safer than glass, they are designed and manufactured for tough applications under rigorous laboratory conditions
Easier to clean than threaded containers or vessels, these one-piece molded, non-threaded carboys are designed for use as receiver or dispensing vessels in biopharmaceutical applications. Solid ported sanitary end caps are also available.
LDPE Carboys
Suited for advanced liquid handling and solution storage for many pharmaceutical and biotechnology applications, these single-use carboys are the ideal choice for storage and transport of reagents. These containers can eliminate cleaning costs, validation and assembly, and contamination from repeated use.    
HDPE Carboys and Jerricans
Manufactured for tough applications, these carboys and jerricans are suited for storing and dispensing solutions and media. We offer a selection of heavy-duty, fluorinated and HDPE jerricans graduated in liters and gallons. 
These Nalgene labware are sterilized and tested after assembly, with sterility guaranteed as long as the packaging is intact. Indicator dots used on Nalgene products undergo an irreversible color change upon sterilization.    
Polypropylene Carboys
Durable, lighter and safer than glass, these carboys are available in a variety of shapes and sizes, with a range of features such as wide mouths and handles for convenient filling and handling
Easier to clean than threaded containers or vessels, these one-piece molded, non-threaded carboys are designed for use as receiver or dispensing vessels in biopharmaceutical applications. Solid ported 
Polycarbonate Carboys
Our polycarbonate carboys ensure the integrity of your samples. Glass-clear to permit easy visual inspection and volume measurements but lighter and safer than glass, they are designed and manufactured for tough applications under rigorous laboratory conditions
Easier to clean than threaded containers or vessels, these one-piece molded, non-threaded carboys are designed for use as receiver or dispensing vessels in biopharmaceutical applications. Solid ported sanitary end caps are also available.
Related products
Polycarbonate Validation Bottles
These bottles of durable polycarbonate construction are transparent and autoclavable, allowing for convenient material compatibility validation.    
Fluid Transfer and Customization
Bring your bioproduction processes together seamlessly with an extensive range of standard and customized fluid transfer products.    
Ordering information
LDPE Carboys
HDPE Carboys and Jerricans
PP Carboys
PC Carboys
Carboy Accessories
Resources
Production Bottles and Accessories
Bioprocessing resources
Click below to access products brochures, training videos, mobile apps, and more.
Support
 Request more info on Bioprocessing