Stainless Steel Support Systems
Large-volume rigid stainless steel outer support containers
The Thermo Scientific HyPerforma Smartainer systems are durable outer support containers made of stainless steel and used in conjunction with single-use BioProcess Containers (BPCs) for storage and in-house liquid handling operations of large-volume, sterile process liquids.
Request information  Download product brochure  
Large-volume stainless steel shipping systems
The Smartainer system, a stainless-steel outer container, is designed to accommodate large-volume BPCs and has the following features and applications:
Key Features:
  • Available in 100, 200, 500, 1,000, 1,500, and 2,000 L
  • 304 stainless steel construction
  • Available in non-jacketed or jacketed options for applications requiring heating and cooling
  • Optional accessories include hoist and carts for mobility and BPC handling
  • Movable by forklift, pallet jack, or optional carts
  • Smooth surface for easy cleaning
  • Standard and custom BPCs a are available for these large volume stainless steel containers
Applications:
  • In-process applications involving handling or storage of sterile process liquids such as:
  • Sterile media and buffer storage and transport within facility
  • Feeding sterile media into bioreactors, and buffers into purification systems
  • Receiving and storage of bioreactor harvest
The Smartainer system uses the following components and associated accessories:
  1. Smartainer or jacketed Smartainer system
    Specially designed stainless steel container to support BPCs
  2. BPC hoist (optional)
    Assists in handling and loading the BPC into the Smartainer system
  3. Tray assembly (optional)
    Conveniently holds dispense pumps, tube welders, and other items
  4. Cart (optional)
    Assists in moving the Smartainer system short distances over smooth and level surfaces
The Smartainer system, a stainless-steel outer container, is designed to accommodate large-volume BPCs and has the following features and applications:
Key Features:
  • Available in 100, 200, 500, 1,000, 1,500, and 2,000 L
  • 304 stainless steel construction
  • Available in non-jacketed or jacketed options for applications requiring heating and cooling
  • Optional accessories include hoist and carts for mobility and BPC handling
  • Movable by forklift, pallet jack, or optional carts
  • Smooth surface for easy cleaning
  • Standard and custom BPCs a are available for these large volume stainless steel containers
Applications:
  • In-process applications involving handling or storage of sterile process liquids such as:
  • Sterile media and buffer storage and transport within facility
  • Feeding sterile media into bioreactors, and buffers into purification systems
  • Receiving and storage of bioreactor harvest
The Smartainer system uses the following components and associated accessories:
  1. Smartainer or jacketed Smartainer system
    Specially designed stainless steel container to support BPCs
  2. BPC hoist (optional)
    Assists in handling and loading the BPC into the Smartainer system
  3. Tray assembly (optional)
    Conveniently holds dispense pumps, tube welders, and other items
  4. Cart (optional)
    Assists in moving the Smartainer system short distances over smooth and level surfaces
Related products
Available in standard or custom configurations in a full range of sizes for a range of applications
Provides you the flexibility to add custom-configured options without extensive lead times or associated costs.
Ordering information
Resources
Storage and transport solutions
BioProduction resources
Click below to access products brochures, training videos, mobile apps, and more.
Support
 Request more info on BioProduction