Request a demo Learn more about EVOS FL Auto
Application guide: Setting up hypoxic conditions with the EVOS onstage incubator Download now
Create stunning time-lapse images in your lab
Designed specifically for the EVOS FL Auto Imaging System, the EVOS Onstage Incubator is an environmental chamber that enables precise control of temperature, humidity, and three gases for time-lapse imaging of live cells under both physiological and non-physiological conditions.
See it in action
Request a demo Learn more about EVOS FL Auto
Application guide: Setting up hypoxic conditions with the EVOS onstage incubator Download now
Create stunning time-lapse images in your lab
Designed specifically for the EVOS FL Auto Imaging System, the EVOS Onstage Incubator is an environmental chamber that enables precise control of temperature, humidity, and three gases for time-lapse imaging of live cells under both physiological and non-physiological conditions.
See it in action
Superb flexibility, ease of use, and optical performance for live cell imaging
Environmental settings and image acquisition parameters are all seamlessly integrated into the EVOS FL Auto interface, creating a high-performance inverted imaging system with superb flexibility, ease of use and optical performance for demanding live cell time-lapse imaging experiments. Features include:
  • Fully integrated environmental chamber for live cell time-lapse imaging
  • Intuitively set environmental and image acquisition parameters
  • Easily maintain physiological or non-physiological conditions with precision control
  • Small footprint, robust build, and sleek design
  • Choice of a range of vessel holders
Time-lapse videos using the EVOS FL Auto with Onstage Incubator
Cell division in HeLa cells
Cells: HeLa
Reagents: CellLight™ H2B GFP, CellLight™ Mito RFP
Time Lapse: 24 hours with images taken every 12 minutes
Imaged on EVOS FL Auto Imaging System using the Onstage Incubator
Disruption of microfilament organization in HeLa cells
Cells: HeLa treated with Latrunculin A
Reagents: CellLight™ Actin GFP, MitoTracker™ Red, NucBlue™ Live
Time Lapse: 14 hours with images taken every 5 minutes
Imaged on EVOS FL Auto Imaging System using the Onstage Incubator
Specifications
Attribute Details
Compatible Vessels Well plates, 35 mm dishes, T25 Flasks
Temperature Range Ambient - 40oC
CO2 % Range 0-20%
O2 % Range Ambient - 0%
Humidity Range >80% RH @ 37oC
Dimensions
  • 25 x 19 x 3.7 cm (Environment chamber)
  • 37 x 16 x 20 cm (Control unit)
Weight
  • 1.5 kg (Environment chamber)
  • 10 kg (Control unit)
EVOS Onstage Incubator resources
EVOS Onstage Incubator user manual
BioProbes™ Journal Article
Scientific Posters
EVOS Accessories
EVOS Application Guides