Revolutionizing fluorescence microscopy
The powerful yet easy-to-use Invitrogen EVOS FL Cell Imaging System provides the flexibility to fit most epifluorescence microscopy applications. Combining precision components with unique design functionality, the EVOS FL Cell Imaging System has revolutionized fluorescence microscopy.
Compare imaging systems ›
Request a demoRequest a quote
EVOS FL demo video
EVOS FL virtual tour
EVOS FL Image Gallery
See our gallery of beautiful images taken with the EVOS FL Cell Imaging System.
EVOS Light Cubes
Find the perfect light cube and reagent combination using our EVOS Light Cube selection guide.
Request an FL Demo
Get the full experience by requesting an in-lab demo from one of our trained representatives.
EVOS imaging systems brochure
See the entire portfolio of easy to use cell imaging instruments that can be configured with a variety of objectives, light cubes, and vessel holders to meet your research needs.
  Download the EVOS brochure
EVOS FL Cell Imaging System Specs
Attribute EVOS FL Detail
Illumination Adjustable-intensity LED (>50,000-hour life per light cube)
Contrast methods Epifluorescence and transmitted light (bright field and phase contrast)
Objective turret 5-position, front-mounted control
Fluorescence channels Simultaneously accommodates up to 4 fluorescent light cubes
Condenser Includes 3 positions, with bright-field and phase-contrast annuli
Condenser working distance 60 mm
Stage Mechanical “glide” stage with X-Y axis fine-positioning controls
Interchangeable vessel holders available
LCD display 15” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt
Cameras
Monochrome:
Sony ICX445 monochrome CCD,
1/3” 1,280 x 960, 1.3 megapixels
Color:
Sony ICX285AQ color CCD,
2/3” 1,360 x 1,024, 1.4 megapixels
Captured images
Monochrome camera:
16-bit monochrome TIFF or PNG (12-bit dynamic range); 24-bit color TIFF or PNG;
JPEG, BMP (1,280 x 960 pixels)
Color camera:
16-bit color TIFF or PNG (12-bit dynamic range); 24-bit color TIFF or PNG;
JPEG, BMP (1,360 x 1,024 pixels)
Output ports 3 USB ports, 1 DVI port (supports direct output to USB and networked storage)
Thermo Fisher Cloud
Save files to Cloud through computer
Power supply AC adaptor
Dimensions Height: 578 mm (22.8 in)
Depth: 470 mm (18.5 in)
Width: 355 mm (14.0 in)
Weight 15.3 kg (33.7 lb)
EVOS FL Cell Imaging System Citations
EVOS FL Cell Imaging System Specs
Attribute EVOS FL Detail
Illumination Adjustable-intensity LED (>50,000-hour life per light cube)
Contrast methods Epifluorescence and transmitted light (bright field and phase contrast)
Objective turret 5-position, front-mounted control
Fluorescence channels Simultaneously accommodates up to 4 fluorescent light cubes
Condenser Includes 3 positions, with bright-field and phase-contrast annuli
Condenser working distance 60 mm
Stage Mechanical “glide” stage with X-Y axis fine-positioning controls
Interchangeable vessel holders available
LCD display 15” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt
Cameras
Monochrome:
Sony ICX445 monochrome CCD,
1/3” 1,280 x 960, 1.3 megapixels
Color:
Sony ICX285AQ color CCD,
2/3” 1,360 x 1,024, 1.4 megapixels
Captured images
Monochrome camera:
16-bit monochrome TIFF or PNG (12-bit dynamic range); 24-bit color TIFF or PNG;
JPEG, BMP (1,280 x 960 pixels)
Color camera:
16-bit color TIFF or PNG (12-bit dynamic range); 24-bit color TIFF or PNG;
JPEG, BMP (1,360 x 1,024 pixels)
Output ports 3 USB ports, 1 DVI port (supports direct output to USB and networked storage)
Thermo Fisher Cloud
Save files to Cloud through computer
Power supply AC adaptor
Dimensions Height: 578 mm (22.8 in)
Depth: 470 mm (18.5 in)
Width: 355 mm (14.0 in)
Weight 15.3 kg (33.7 lb)
EVOS FL Cell Imaging System Citations
EVOS FL Application Videos
Minimizing photobleaching
Cell Counting
Choosing proper objectives
Resources
EVOS FL Software Update
EVOS FL User Manual
EVOS FL Quick Start Guide 
EVOS Accessories
Webinars
Cell Analysis Support Center
Find tips, troubleshooting help, and resources for your cell analysis workflow