EVOS XL Core Cell Imaging System
Easy cell monitoring and imaging in your cell culture hood
The Invitrogen EVOS XL Core Cell Imaging System is the perfect addition to any cell culture room or facility. Ergonomically designed, the all-in-one XL Core system can fit into your tissue culture hood or on an adjacent bench, making it easy for you to quickly analyze the status of your cell cultures.
Request a demo    Request a quote
Compare imaging instruments
See the gallery of beautiful images taken with the Invitrogen EVOS XL Core Cell Imaging System.
See what your peers are publishing with all the Invitrogen EVOS Cell Imaging Systems.
Get the full experience by requesting an in-lab demo from one of our trained representatives.
EVOS imaging systems brochure
See the entire portfolio of easy to use cell imaging instruments that can be configured with a variety of objectives, light cubes, and vessel holders to meet your research needs.
Download the EVOS brochure
EVOS XL Core application videos
Viewing Images 
Capturing and Saving Images 
Manual and On-Screen Controls 
System Highlights
Attribute EVOS XL Core Detail
Optics Infinity-corrected optical system; RMS-threaded objectives with 45 mm parfocal distance
Illumination Adjustable-intensity LED (>50,000-hour life per light cube)
Contrast methods Transmitted light (bright field and phase contrast)
Objectives 4x, 10x, 20x, 40x; plan LWD phase, infinity;
Optional 2x (no phase)
Objective turret 4-position, manual control
Condenser 3-position turret for bright field and phase contrast
Condenser working distance 60 mm
Stages
Fixed stage (AMEX1000);
Optional mechanical stage (AMEP4712):
  • X-Y axis controls and vessel holder framework
  • 120 mm x 78 mm range of motion
  • Both knobs 56 mm/rev
Focus mechanism Coaxial focus knobs with tension control:
  • Coarse focus: 38 mm/rev
  • Fine focus: 0.2 mm/rev, precision 0.002 mm
LCD display 12.1” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt
Camera Color: 1/2” 2,048 x 1,536, 3.1 megapixels
Image acquisition Embedded operating system with imaging software for image capture and save via mouse or front-mounted manual buttons
Captured images Color camera: 24-bit full-color TIFF or PNG; JPEG, BMP (2,048 x 1,536 pixels)
Output ports 2 USB 2.0 ports
Power supply AC adaptor; input 100–240 V, 47–63 Hz; output 12 V DC/2.0 A 24 W max
Dimensions Height: 533 mm (21.0 in)
Depth: 406 mm (6.0 in)
Width: 318 mm (12.5 in)
Weight 9.1 kg (20.1 lb);
10.0 kg (22.1 lb) with mechanical stage
EVOS XL Core Citations
Thermo Fisher Cloud provides scientists with a secure place to store, share and analyze their data. Upload your files from anywhere you have access to the Internet by simply dragging them to the Cloud folder on your Windows™ desktop. Share the results with your colleagues to observe patterns, validate findings, and develop insights. And it’s not just for data files. Upload a presentation for an upcoming conference or a particularly useful how-to video.  
Get started today with 10 GB of free storage.
Learn more ›
Access it now › 
Use your existing web account username and password or register 
System Highlights
Attribute EVOS XL Core Detail
Optics Infinity-corrected optical system; RMS-threaded objectives with 45 mm parfocal distance
Illumination Adjustable-intensity LED (>50,000-hour life per light cube)
Contrast methods Transmitted light (bright field and phase contrast)
Objectives 4x, 10x, 20x, 40x; plan LWD phase, infinity;
Optional 2x (no phase)
Objective turret 4-position, manual control
Condenser 3-position turret for bright field and phase contrast
Condenser working distance 60 mm
Stages
Fixed stage (AMEX1000);
Optional mechanical stage (AMEP4712):
  • X-Y axis controls and vessel holder framework
  • 120 mm x 78 mm range of motion
  • Both knobs 56 mm/rev
Focus mechanism Coaxial focus knobs with tension control:
  • Coarse focus: 38 mm/rev
  • Fine focus: 0.2 mm/rev, precision 0.002 mm
LCD display 12.1” high-resolution (1,024 x 768 pixels) color monitor with adjustable tilt
Camera Color: 1/2” 2,048 x 1,536, 3.1 megapixels
Image acquisition Embedded operating system with imaging software for image capture and save via mouse or front-mounted manual buttons
Captured images Color camera: 24-bit full-color TIFF or PNG; JPEG, BMP (2,048 x 1,536 pixels)
Output ports 2 USB 2.0 ports
Power supply AC adaptor; input 100–240 V, 47–63 Hz; output 12 V DC/2.0 A 24 W max
Dimensions Height: 533 mm (21.0 in)
Depth: 406 mm (6.0 in)
Width: 318 mm (12.5 in)
Weight 9.1 kg (20.1 lb);
10.0 kg (22.1 lb) with mechanical stage
EVOS XL Core Citations
Thermo Fisher Cloud provides scientists with a secure place to store, share and analyze their data. Upload your files from anywhere you have access to the Internet by simply dragging them to the Cloud folder on your Windows™ desktop. Share the results with your colleagues to observe patterns, validate findings, and develop insights. And it’s not just for data files. Upload a presentation for an upcoming conference or a particularly useful how-to video.  
Get started today with 10 GB of free storage.
Learn more ›
Access it now › 
Use your existing web account username and password or register 
Resources
EVOS XL Core Software Update
Download the latest EVOS XL Core Software
EVOS XL Core User Manual
 Download the EVOS XL Core user manual [PDF, 1.8 MB]
EVOS XL Core Quick Start Guide
 Download the EVOS XL Core Quick Start Guide [PDF, 2.3 MB] 
EVOS Accessories
EVOS objectives
Webinars
From the hood to the microscope: Revolutionizing cell-based imaging
Cell Analysis Support Center
Find tips, troubleshooting help, and resources for your cell analysis workflow.