Image-iT® FX Kits  
All-in-one kits for fluorescence imaging
The Image-iT® FX Kits provide the dyes and reagents you need for optimal immunofluorescence imaging of fixed cells and tissue sections. Each kit includes:
A pack of two CultureWell® chambered coverglasses is also included for convenient processing. Each Image-iT® FX Kit provides sufficient materials to perform 50–100 assays.
View all products
Alexa Fluor® secondary detection conjugates
The Alexa Fluor® dye conjugates are, without exception, the best conjugates we have ever tested. With excellent brightness and photostability, these fluorescent conjugates of streptavidin, goat anti–mouse, and goat anti–rabbit IgG are superior fluorescent secondary reagents for imaging.
Alexa Fluor® secondary detection conjugates  
Labeling cells with the bright and photostable Alexa Fluor® dye conjugates yields stunning, high-quality images.
ProLong® Gold antifade reagent
ProLong® Gold antifade reagent is premixed and ready to use—just add a drop to your preparation and mount. ProLong® Gold reagent outperforms most other commercially available antifade reagents, significantly reducing fluorophore photobleaching while causing little or no quenching of the fluorescence signal. This reagent offers excellent compatibility with a multitude of dyes and dye complexes, making it an especially valuable tool for multicolor applications. ProLong® Gold reagent cures within 24 hours, and the sample can be saved for months after mounting.
ProLong® Gold antifade reagent
ProLong® Gold antifade reagent (top) provides significant photobleaching protection compared to PBS alone (bottom) over a 20-second time series. Images were acquired at 5-second intervals.
Image-iT® FX signal enhancer
Image-iT® FX signal enhancer dramatically improves the signal-to-noise ratio of immunolabeled cells and tissues, allowing you to clearly visualize targets that would normally be indistinguishable due to background fluorescence. Background staining typically seen with fluorescent dyes is largely eliminated when Image-iT® FX signal enhancer is applied to fixed and permeabilized cells before staining (Table 1).
Image-iT® FX signal enhancer  
Treatment with Image-iT® FX signal enhancer
(left panels) largely eliminates nonspecific fluorescent dye binding (right panels) in (A) cultured cells, and (B) a tissue section.
Table 1. Fluorescent dyes successfully tested with the Image-iT® FX Signal Enhancer.*
Alexa Fluor® 350 Alexa Fluor® 633 Cascade Blue®
Alexa Fluor® 405 Alexa Fluor® 635 Pacific Blue™
Alexa Fluor® 430 Alexa Fluor® 647 Fluorescein
Alexa Fluor® 488
Alexa Fluor® 660 Oregon Green® 488
Alexa Fluor® 532 Alexa Fluor® 680 Tetramethylrhodamine
Alexa Fluor® 546 Alexa Fluor® 700 Cy®3
Alexa Fluor® 555 Alexa Fluor® 750 Rhodamine B
Alexa Fluor® 568   Rhodamine Red-X
Alexa Fluor® 594   Texas Red®, Texas Red-X
Alexa Fluor® 610   Cy®5
* All dyes were conjugated to streptavidin and tested at 10 µg/mL.
Resources
More information
Application areas