2018 Attune NxT Flow Cytometer
Efficient. Flexible. Transformative.
Configure your Attune NxT Flow Cytometer with fully automated sample acquisition from 96-well and 384-well microtiter plates with the Invitrogen Attune Autosampler.
Advantages of the Attune NxT Flow Cytometer Automation Configuration include:
  • Compatibility—Attune Autosampler is compact and compatible with many different types of plates
  • Volume—No carousel needed; run deep well plates up to 2 mL
  • One-click transition—no disassembly, no additional QC, no reboot required for conversion between plates and tubes
Request a demoRequest a quote
Attune Autosampler
  • Broad compatibility—Compatible with many different plate formats, including 96-well, 384-well, and deep-well plates.
  • Intelligent probe—Intelligent probe design minimizes clogging and carryover (1.0%) and prevents damage to the instrument.
  • Auto cleaning—Performs automated cleaning when the instrument is shutting down.
  • Consistent data—Designed to provide minimal variation regardless of sampling method (tube vs. plate) and collection rate.
  • Mixing by aspiration—Mixing sample by aspiration instead of shaking ensures homogeneity of the sample and maintains cell viability. See why mixing sample by aspiration is better ›
Attune NxT Flow Cytometer Automation Configuration
Attune NxT Flow Cytometer
Attune NxT Flow Cytometer
  • Efficient—up to 10x faster than traditional flow cytometric analyzers
  • Flexible—up to 14 colors and up to 6 channels with the violet laser
  • Transformative—run difficult samples with a system that has active mechanical resistance to clogging
Learn more about the Attune NxT Flow Cytometer
Attune NxT Autosampler
Attune Autosampler
  • Low carryover—standard mode <0.5% (two wash cycles); multiple-rinse capability for ultralow carryover
  • Thorough sample mixing—each well is mixed via full aspiration (no shaking) to preserve cell viability
  • Maximized sampling—small dead volume of 30 µL for 12.5–200 µL/min, 50 µL for 50–1,000 µL/min
Request a demoRequest a quote
Fluids consumables
Extended fluidics option
The Attune NxT Flow Cytometer Automation Configuration is available with the Attune NxT External Fluid Supply (EFS), with 10 L fluid capacity to support extended runtimes. The EFS supplements the on-board fluidics of the Attune NxT Flow Cytometer.
Attune NxT External Fluid Supply (EFS) offers:
  • Extended fluid options—a 10 L cube increases the period of continual operation by reducing the frequency of replenishing and emptying the fluid
  • Active monitoring—waste bottle features a vent, sensor, and fill-line alarm
  • Benchtop flexibility—umbilical cords enable storage either under the bench or on an adjacent cart
Specifications for the Attune Autosampler
Performance
  • High-throughput mode acquisition time: <42 min for 96-well plate, <180 min for 384-well plate (using one rinse and one mix, and full analysis of a 40 μL sample)
  • Carryover: <0.5% in plate loader format (standard mode, 2 wash cycles); multiple-rinse capability for ultralow carryover
  • Sample mixing: Mixing optimized to preserve cell viability; mixing cycles optimized to sample analysis volume
  • Mixing method: Each well mixed via full aspiration (no shaking)
  • Wash cycle: User-defined number of wash cycles, dependent on plate-processing protocol and time to acquire plates
  • Minimum dead volume (single draw): 30 μL for 12.5–200 μL/min, 50 μL for 50–1,000 μL/min
  • Sample window: Protectively coated window allows visibility to well progress while preventing exposure to ambient light during acquisition
  • Auto-calibration: Regular, 30-day interval, system-initiated function
Fluidics
  • Plate and tube compatibility: One-click transition from tubes to plates and vice versa; no disassembly, no additional QC, no reboot required for conversion between plates and tubes
  • Compatible plate types: 96 deep-well (flat, round, and V-bottom), 96-well standard depth (flat, round, and V-bottom), 384-well standard depth (flat, round, and V-bottom), 384 deep-well (flat, round, and V-bottom)
  • Cleaning cycles: Automated daily and monthly cleaning protocols
  • Fluidics requirements: 800 mL total of onboard fluid tanks, capable of running four 96-well plates
  • Extended fluidics option: Optional external fluid tank with 10 L fluid capacity
Installation requirements
  • Size (W x D x H): ~40 x 29 x 29 cm (16 x 11 x 11 in.)
  • Space requirements:
    • Minimum width: 40 cm (15.8 in.); when attached to the Attune NxT Flow Cytometer, the total width is 167 cm (65.8 in.)
    • Minimum depth: 58.5 cm (23.1 in.) provides 43.2 cm (17.1 in.) for the cytometer unit, a 10.2 cm (4 in.) ledge in front of the unit to place fluidics bottles, and 6.5 cm (2.5 in.) behind the unit for ventilation
    • Minimum clear height: 74 cm (29 in.) above the mounting
  • Mounting: Side
  • Weight: ~16 kg (35 lb)
  • Operating range (environmental conditions): 15–30°C (50–95°F)
  • Operating humidity: <80% noncondensing
  • Electrical requirements: 100–240 VAC, 50/60 Hz, <300 W
Designed to minimize clogging and carryover
The Attune Autosampler, with intelligent probe design, is designed to minimize clogging and carryover (1.0%) (Table 1) and prevent damage to the instrument. Mixing by gentle aspiration helps to ensure sample homogeneity and to maintain cell viability (Table 2).
Table 1. Minimal carryover using the Attune NxT Autosampler. Jurkat cells at a concentration of 1 x 106 cells/mL were dispensed into a 96-well, v-bottom plate, sampled using the Attune NxT Autosampler, and analyzed on the Attune NxT flow cytometer at two sample rates. Percent sample carryover was calculated for each wash routine.
  Number of washes and % carryover
  1 wash
2 washes
3 washes
Standard mode 0.01 0.01 0.01
High throughput mode
0.02 0.02 0.02
Table 2. Increasing the number of mixing cycles on the Attune NxT Autosampler does not adversely affect cell viability. Ammonium chloride–lysed whole blood (LWB) and NIH/3T3 (live/heat-treated) cells were stained with 2 μg/mL propidium iodide (a dead-cell indicator), loaded in a 96-well plate (in triplicate), mixed 0–5 times using the Attune NxT Autosampler, and analyzed on the Attune NxT flow cytometer.
  Percent dead cells (%)
Number of mix cycles LWB NIH/3T3
0
0.75 34.10
1
0.78 32.83
2
0.74 33.52
3
0.74 32.75
4
0.74 33.26
5
0.75 31.58
Straightforward run set-up
Setting up a multiwell plate sampling run using the Attune NxT software is simple. Using a virtual plate layout view, you can create compensation wells, define instrument settings, and identify samples. The software also allows you to define multiple experiments on a single plate and recover unused sample (for samples collected or acquired in tube mode only).
The intuitive software is easy to use and enables quick data analysis. For example, a heat-map view allows rapid screening and confirmation of samples while the instrument is acquiring data (Figure 1).
flow cytometer heat map
Figure 1. Attune NxT Autosampler heat map function identifies variation within a parameter across a 96-well plate. Live and heat-killed THP-1 cells were stained with propidium iodide and dispensed into a 96-well plate. Samples were applied to the Attune NxT flow cytometer using the Attune Nxt Autosampler. Heat map (indigo correlating with live cells and red with dead cells) and numeric overlay text provide real-time feedback on the assay being performed.
Compatible with a range of multiwell plates
The Attune Autosampler is compatible with a range of shallow and deep-well, 96 and 384 well plates.
Compatible plate types
  • 96 deep-well (flat, round, and V-bottom)
  • 96-well standard depth (flat, round, and V-bottom)
  • 384-well standard depth (flat, round, and V-bottom)
  • 384 deep-well (flat, round, and V-bottom)
Plate/tube compatibility
One-click transition from tubes to plates and vice versa. No disassembly, additional QC or reboot required for conversion between plate and tube.
Reliable utility for diverse applications
Automation helps maximize reproducibility between experiments and reduces inter-assay variability and inter-operator variability. By demonstrating low CVs across and through plates, the Attune NxT Flow Cytometer with Autosampler offers a reliable and high-throughput approach for multiparametric cell screening with 96- and 384-well plates, allowing for less sample quantities to maximize the efficient use of a limited numbers of cells. This combination of higher-throughput with higher-content flow cytometric analysis of as many as 16 parameters for each cell with acquisition speeds of up to 1 mL/min provides a powerful screening platform for drug discovery and systems biology experiments. See some examples of applications enabled by Attune NxT Flow Cytometer with Autosampler below.
Flow Cytometry Analysis of Dose Response for Apoptosis Induction
Read the application note
Live/Dead Algal Analysis With the Attune NxT Flow Cytometer
Read the application note
High-Throughput Automation With the Attune NxT Autosampler: Consistent Results Across All Wells and Across Plates
Read the application note
Consistent results regardless of sampling method
The Attune NxT Flow Cytometer Automation configuration with the Attune Autosampler demonstrates consistent results regardless of whether the sample is delivered from a tube or from a plate (Figure 3).
Comparison of scatter plots  obtained from a plate and a tube show minimal variation
Figure 3. Attune NxT Autosampler offers consistent results regardless of sampling method. Lysed whole blood was stained for CD45, CD4, and CD8 markers, aliquotted into both a tube and a plate, and analyzed on an Attune NxT flow cytometer. Minimal variation was observed between the sampling modes.
Attune Autosampler
  • Broad compatibility—Compatible with many different plate formats, including 96-well, 384-well, and deep-well plates.
  • Intelligent probe—Intelligent probe design minimizes clogging and carryover (1.0%) and prevents damage to the instrument.
  • Auto cleaning—Performs automated cleaning when the instrument is shutting down.
  • Consistent data—Designed to provide minimal variation regardless of sampling method (tube vs. plate) and collection rate.
  • Mixing by aspiration—Mixing sample by aspiration instead of shaking ensures homogeneity of the sample and maintains cell viability. See why mixing sample by aspiration is better ›
Attune NxT Flow Cytometer Automation Configuration
Attune NxT Flow Cytometer
Attune NxT Flow Cytometer
  • Efficient—up to 10x faster than traditional flow cytometric analyzers
  • Flexible—up to 14 colors and up to 6 channels with the violet laser
  • Transformative—run difficult samples with a system that has active mechanical resistance to clogging
Learn more about the Attune NxT Flow Cytometer
Attune NxT Autosampler
Attune Autosampler
  • Low carryover—standard mode <0.5% (two wash cycles); multiple-rinse capability for ultralow carryover
  • Thorough sample mixing—each well is mixed via full aspiration (no shaking) to preserve cell viability
  • Maximized sampling—small dead volume of 30 µL for 12.5–200 µL/min, 50 µL for 50–1,000 µL/min
Request a demoRequest a quote
Fluids consumables
Extended fluidics option
The Attune NxT Flow Cytometer Automation Configuration is available with the Attune NxT External Fluid Supply (EFS), with 10 L fluid capacity to support extended runtimes. The EFS supplements the on-board fluidics of the Attune NxT Flow Cytometer.
Attune NxT External Fluid Supply (EFS) offers:
  • Extended fluid options—a 10 L cube increases the period of continual operation by reducing the frequency of replenishing and emptying the fluid
  • Active monitoring—waste bottle features a vent, sensor, and fill-line alarm
  • Benchtop flexibility—umbilical cords enable storage either under the bench or on an adjacent cart
Specifications for the Attune Autosampler
Performance
  • High-throughput mode acquisition time: <42 min for 96-well plate, <180 min for 384-well plate (using one rinse and one mix, and full analysis of a 40 μL sample)
  • Carryover: <0.5% in plate loader format (standard mode, 2 wash cycles); multiple-rinse capability for ultralow carryover
  • Sample mixing: Mixing optimized to preserve cell viability; mixing cycles optimized to sample analysis volume
  • Mixing method: Each well mixed via full aspiration (no shaking)
  • Wash cycle: User-defined number of wash cycles, dependent on plate-processing protocol and time to acquire plates
  • Minimum dead volume (single draw): 30 μL for 12.5–200 μL/min, 50 μL for 50–1,000 μL/min
  • Sample window: Protectively coated window allows visibility to well progress while preventing exposure to ambient light during acquisition
  • Auto-calibration: Regular, 30-day interval, system-initiated function
Fluidics
  • Plate and tube compatibility: One-click transition from tubes to plates and vice versa; no disassembly, no additional QC, no reboot required for conversion between plates and tubes
  • Compatible plate types: 96 deep-well (flat, round, and V-bottom), 96-well standard depth (flat, round, and V-bottom), 384-well standard depth (flat, round, and V-bottom), 384 deep-well (flat, round, and V-bottom)
  • Cleaning cycles: Automated daily and monthly cleaning protocols
  • Fluidics requirements: 800 mL total of onboard fluid tanks, capable of running four 96-well plates
  • Extended fluidics option: Optional external fluid tank with 10 L fluid capacity
Installation requirements
  • Size (W x D x H): ~40 x 29 x 29 cm (16 x 11 x 11 in.)
  • Space requirements:
    • Minimum width: 40 cm (15.8 in.); when attached to the Attune NxT Flow Cytometer, the total width is 167 cm (65.8 in.)
    • Minimum depth: 58.5 cm (23.1 in.) provides 43.2 cm (17.1 in.) for the cytometer unit, a 10.2 cm (4 in.) ledge in front of the unit to place fluidics bottles, and 6.5 cm (2.5 in.) behind the unit for ventilation
    • Minimum clear height: 74 cm (29 in.) above the mounting
  • Mounting: Side
  • Weight: ~16 kg (35 lb)
  • Operating range (environmental conditions): 15–30°C (50–95°F)
  • Operating humidity: <80% noncondensing
  • Electrical requirements: 100–240 VAC, 50/60 Hz, <300 W
Designed to minimize clogging and carryover
The Attune Autosampler, with intelligent probe design, is designed to minimize clogging and carryover (1.0%) (Table 1) and prevent damage to the instrument. Mixing by gentle aspiration helps to ensure sample homogeneity and to maintain cell viability (Table 2).
Table 1. Minimal carryover using the Attune NxT Autosampler. Jurkat cells at a concentration of 1 x 106 cells/mL were dispensed into a 96-well, v-bottom plate, sampled using the Attune NxT Autosampler, and analyzed on the Attune NxT flow cytometer at two sample rates. Percent sample carryover was calculated for each wash routine.
  Number of washes and % carryover
  1 wash
2 washes
3 washes
Standard mode 0.01 0.01 0.01
High throughput mode
0.02 0.02 0.02
Table 2. Increasing the number of mixing cycles on the Attune NxT Autosampler does not adversely affect cell viability. Ammonium chloride–lysed whole blood (LWB) and NIH/3T3 (live/heat-treated) cells were stained with 2 μg/mL propidium iodide (a dead-cell indicator), loaded in a 96-well plate (in triplicate), mixed 0–5 times using the Attune NxT Autosampler, and analyzed on the Attune NxT flow cytometer.
  Percent dead cells (%)
Number of mix cycles LWB NIH/3T3
0
0.75 34.10
1
0.78 32.83
2
0.74 33.52
3
0.74 32.75
4
0.74 33.26
5
0.75 31.58
Straightforward run set-up
Setting up a multiwell plate sampling run using the Attune NxT software is simple. Using a virtual plate layout view, you can create compensation wells, define instrument settings, and identify samples. The software also allows you to define multiple experiments on a single plate and recover unused sample (for samples collected or acquired in tube mode only).
The intuitive software is easy to use and enables quick data analysis. For example, a heat-map view allows rapid screening and confirmation of samples while the instrument is acquiring data (Figure 1).
flow cytometer heat map
Figure 1. Attune NxT Autosampler heat map function identifies variation within a parameter across a 96-well plate. Live and heat-killed THP-1 cells were stained with propidium iodide and dispensed into a 96-well plate. Samples were applied to the Attune NxT flow cytometer using the Attune Nxt Autosampler. Heat map (indigo correlating with live cells and red with dead cells) and numeric overlay text provide real-time feedback on the assay being performed.
Compatible with a range of multiwell plates
The Attune Autosampler is compatible with a range of shallow and deep-well, 96 and 384 well plates.
Compatible plate types
  • 96 deep-well (flat, round, and V-bottom)
  • 96-well standard depth (flat, round, and V-bottom)
  • 384-well standard depth (flat, round, and V-bottom)
  • 384 deep-well (flat, round, and V-bottom)
Plate/tube compatibility
One-click transition from tubes to plates and vice versa. No disassembly, additional QC or reboot required for conversion between plate and tube.
Reliable utility for diverse applications
Automation helps maximize reproducibility between experiments and reduces inter-assay variability and inter-operator variability. By demonstrating low CVs across and through plates, the Attune NxT Flow Cytometer with Autosampler offers a reliable and high-throughput approach for multiparametric cell screening with 96- and 384-well plates, allowing for less sample quantities to maximize the efficient use of a limited numbers of cells. This combination of higher-throughput with higher-content flow cytometric analysis of as many as 16 parameters for each cell with acquisition speeds of up to 1 mL/min provides a powerful screening platform for drug discovery and systems biology experiments. See some examples of applications enabled by Attune NxT Flow Cytometer with Autosampler below.
Flow Cytometry Analysis of Dose Response for Apoptosis Induction
Read the application note
Live/Dead Algal Analysis With the Attune NxT Flow Cytometer
Read the application note
High-Throughput Automation With the Attune NxT Autosampler: Consistent Results Across All Wells and Across Plates
Read the application note
Consistent results regardless of sampling method
The Attune NxT Flow Cytometer Automation configuration with the Attune Autosampler demonstrates consistent results regardless of whether the sample is delivered from a tube or from a plate (Figure 3).
Comparison of scatter plots  obtained from a plate and a tube show minimal variation
Figure 3. Attune NxT Autosampler offers consistent results regardless of sampling method. Lysed whole blood was stained for CD45, CD4, and CD8 markers, aliquotted into both a tube and a plate, and analyzed on an Attune NxT flow cytometer. Minimal variation was observed between the sampling modes.
3D Demo: Explore the Attune NxT Flow Cytometer and Autosampler
A software tool for exploring the hardware, software, optics and fluidics of the Attune NxT Flow Cytometer. Hot spots throughout the tool feature data, researcher opinions, and technical specifications. Mouse operation enables rotation, zoom, and measurement. Available in 8 languages.
Design your flow panels using the Invitrogen Flow Cytometer Panel Builder with advanced functionality.
thermofisher.com/flowpanel
Learn what to consider when compare flow cytometers made by multiple manufacturers.
thermofisher.com/compareflow
Resources
User guides and publications
Product selection
Support