Cell Cycle Analysis in Fixed Cells
Multicolor cell cycle studies are possible using flow cytometry and it is advantageous to analyze DNA content on alternative lasers to preserve the common 488 nm laser for other markers. This flexibility can be applied to cell cycle analysis using FxCycle Violet, SYTOX AADvanced, FxCycle PI/RNAse Solution, or FxCycle Far Red stains on the violet, blue, or red lasers, respectively. These products:
- Allow increased flexibility for multicolor cell cycle studies
- Require little or no compensation with 488 nm excitable dyes
- Result in tight CVs for more accurate analysis
Violet laser—fixed cell cycle analysis
FxCycle Violet
FxCycle Violet Stain is a violet laser–excited dye used for cell cycle analysis in fixed cells. Using FxCycle Violet for cell cycle analysis increases the ability to multiplex by freeing up the 488 nm and 633 nm lasers for other cellular analyses such as immunophenotyping, apoptosis analysis, and dead cell discrimination.
Because of its narrow emission spectra, FxCycle Violet Stain overlaps less with other fluorescent channels than other commonly used dyes (propidium iodide and 7-AAD), resulting in minimal compensation requirements and more accurate data.
Blue laser—fixed cell cycle analysis
SYTOX AADvanced Dead Cell Stain
SYTOX AADvanced Dead Cell Stain is a new high-affinity nucleic acid stain for the detection of dead cells and analysis of cell cycle using the common 488 nm blue laser in flow cytometry. The dye is spectrally similar to 7-AAD but with rapid kinetics of uptake and relatively low CVs.
SYTOX AADvanced Dead Cell Stain penetrates cells more efficiently than 7-AAD providing better separation of live and dead cells. SYTOX AADvanced Dead Cell Stain can also be used with fixed cells for DNA content analysis when paired with RNAse treatment. SYTOX AADvanced Dead Cell Stain Kits contain a vial(s) of dried dye and anhydrous DMSO providing stable shelf life.
FxCycle PI/RNAse Solution
FxCycle PI/RNase Staining Solution is used for flow cyotmetric analysis of DNA content in fixed cells. Propidium iodide (PI), a popular red-fluorescent stain, binds to DNA by intercalating between the bases with little or no sequence preference and with a stoichiometry of one dye per 4–5 base pairs of DNA. PI also binds to RNA, necessitating treatment with RNase to distinguish between RNA and DNA staining. This propidium iodide solution includes DNase-free RNase A and a permeablization reagent. It comes ready to use: simply add the staining solution to fixed cells, incubate, and acquire on a flow cytometer without washing.
Red laser—fixed cell cycle analysis
FxCycle Far Red
FxCycle Far Red stain is used for flow cytometric analysis of DNA content in fixed cells when combined with RNAse. This dye takes advantage of the commonly available 633⁄5 nm excitation sources with emission around 660 nm.
With multicolor cell cycle studies possible using flow cytometry, it is often necessary to analyze DNA content on alternative lasers to preserve the common 488 nm laser and detection channels for other markers. Well suited for the popular red laser, FxCycle Far Red stain is a good choice for DNA content analysis in multicolor cell cycle studies.
Resources
Flow Cytometry Learning Center—Access flow cytometry educational resources for better experiment planning and execution.
Fluorescence SpectraViewer—Assess the spectral compatibility of flurophores.
Support
Flow Cytometry Support Center—Find technical support recommendations for your flow cytometry workflows, including tips for experimental setup and in-depth troubleshooting help.
For Research Use Only. Not for use in diagnostic procedures.