Flow Cytometry Beads for Instrument Controls & Standards
Consistent instrument performance, minimal data variation
  • Alignment, size calibration, and sorting beads for optimal instrument performance
  • Compensation & counting beads and live/dead assays for accurate data
  • Compatible with any instrument
Flow cytometers are designed to perform quantitative measurements on individual cells and other particles with high precision, speed, and accuracy, making instrument calibration a critical step in flow cytometer performance and data integrity. Experimental controls for the analysis of fluorescently labeled cells include compensation, absolute cell counts, and live vs. dead determination.
Our flow cytometer alignment beads, cell sorting set-up beads, and size calibration and reference kits ensure that your flow cytometer and experimental design are tuned for maximal performance to provide accurate data collection and analysis.
Ensure the integrity of your experimental data with bead-based compensation controls.
Find tools for cell concentration and absolute counting, as well as distinguishing and quantitating live and dead bacteria.
Resources
Flow Cytometry Learning Center—Access flow cytometry educational resources for better experiment planning and execution.
Flow Cytometry Panel Builderbeta—Design your flow cytometry panel with this online tool for a simplified, customizable experience to fit your needs.
Support
Flow Cytometry Support Center—Find technical support recommendations for your flow cytometry workflows, including tips for experimental setup and in-depth troubleshooting help.
Flow Cytometry Protocols
Contact us