Luminex® Instruments
Luminex® systems are built on proven technology—flow cytometry, microspheres, lasers, digital signal processing, and traditional chemistry—combined in a unique way. Featuring a flexible, open-architecture design, Luminex® xMAP® technology can perform a wide variety of bioassays quickly, cost-effectively, and accurately.
MAGPIX® System
MAGPIX® System
The compact, versatile and affordable way to multiplex. Introducing the MAGPIX® System.
Luminex® 200™ System
The Luminex® 200™ System
Simultaneously quantify multiple proteins in a single well of a microtiter plate.
FLEXMAP 3D® System
FLEXMAP 3D® System
This innovative instrument design takes bead-based multiplexing technology to the next dimension.
xMAP® Technology
xPONENT® software is a modular, flexible, software package for control of the MAGPIX®, Luminex®100/200™, and FLEXMAP 3D® systems.
  • Intuitive graphical user interface for simplified workflow
  • Automated startup, shutdown, and routine maintenance operations
  • Enhanced data analysis and report generation capabilities
  • 21 CFR Part 11 ready
  • Automation and LIS connectivity
Learn more about xPONENT® Software
Luminex® Instrument Specifications
Affordable, efficient and compact in size
Versatile, efficient, and the standard in multiplexing
High throughput, high plex, automation compatible
MAGPIX® System
Luminex® 200™ System
FLEXMAP 3D® System
Optics
LED/CCD Camera
Lasers/APDs/PMTs
Lasers/APDs/PMTs
Hardware
Fluorescent Imager
Flow Cytometry based
Flow Cytometry based
Bead Compatibility
Magnetic
Magnetic/Polystyrene
Magnetic/Polystyrene
Multiplex Capacity
50
100
500
Read Time (96-well plate)
~ 60min
~ 40 min
~ 20 min
Dynamic Range
3.5 logs
3.5 logs
4.5 logs
Microtiter Plate
96 well
96 well
96 and 384 well
Resources
FLEXMAP 3D® System
FLEXMAP 3D® System
This innovative instrument design takes bead-based multiplexing technology to the next dimension.