Luminex 200 System
The Luminex® 200™ System is a compact analyzer that performs up to 100 assays simultaneously in a single well of a microtiter plate. This flexible system is based on the principles of flow cytometry, integrating key xMAP® detection components to meet the multiplex testing needs of clinical and research laboratory professionals.
The Total Luminex® 200™ System includes:
  • Luminex® 200™ Analyzer
  • Luminex® XYP™ Platform
  • Luminex® SDS™ (Sheath Delivery System)
  • xPONENT® v3.1 Software
  • Dell PC, 17” LCD flat screen monitor, keyboard, and mouse
  • Calibration Kit
  • Verification Kit
  • Automated Maintenance Plate
  • Sheath Fluid
  • One year Gold Warranty
  • Installation
  • Full on-site Training