APEX™ Antibody Labeling KitsZenon® Antibody Labeling Kits
APEX­™ Antibody Labeling Kits
APEX™ Antibody Labeling Kits
Covalently label as little as 10–20 µg of IgG primary antibody and compatible with stabilizing proteins such as BSA.
Zenon® Antibody Labeling Kits
Zenon® Antibody Labeling Kits
Label as little as 1 µg  of  IgG primary antibody without losing antibody during the labeling.
More Antibody Labeling Kits
Antibody Labeling from A to Z
Your solutions for conveniently labeling 1 µg or 1 mg antibody, covalently or noncovalently.
  • Learn more about Antibody Labeling from A to Z