Gibco DMEM delivers superior quality, consistency and control for your mammalian cell culture.
Choose the right DMEM
Use the Media Formulation Tool or select below.
Healthier cells live longer
GlutaMAX media are standard cell culture media that contain a stabilized form of L-glutamine, the dipeptide L-alanyl-L-glutamine, that prevents degradation and ammonia buildup in both adherent and suspension cultures. Use GlutaMAX media to increase media stability, minimize toxic ammonia buildup, and maximize cell performance. Get the most from your cell cultures with GlutaMAX media.
Learn more about:
GlutaMAX media ›   GlutaMAX-I supplements ›
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products