Cell-Specific Transfection Protocols
Note: Our new Lipofectamine® Reagent Protocols have been newly optimized for efficiency, viability, and reproducibility across a broad range of cell types. This is often the best place to start, especially in a new cell line. If you find this doesn’t work for your specific cell type, then you can you look to our cell specific protocols below for further optimization.
Cell Type | Source | Application | Recommended Transfection Reagent | Recommended Transfection SKU | Protocol/ Reference |
||
---|---|---|---|---|---|---|---|
0 - 9 | |||||||
10T1/2 | Mouse embryonic fibroblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
293-F | Human kidney | Protein Production | 293fectin™ | 12347-019 | ![]() |
||
293T | Human embryonic kidney | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
3T3L1 | Mouse fibroblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
3T3L1 | Mouse fibroblast | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
786-O | Human renal cell adenocarcinoma cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
A-F | |||||||
A549 | Human lung carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
A549 | Human lung carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
ACHN | Human renal cancer | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
ARPE-19 | Human retina | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
B16-F10 | Mouse melanoma | Plasmid DNA | Lipofectamine® | 18324-012 | 18324-020 | ![]() |
|
BAEC | Bovine aortic endothelial cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
BGOV-1 | Human embryonic stem cells | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
BHK-21 | Hamster kidney | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Breast Adipose Fibroblasts | Human breast adipose | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
C2C12 | Mouse muscle myoblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
C3A | Human hepatocellular Carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
C6 | Rat glioma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Caco-2 | Human colorectal adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Cardiac Fibroblasts | Adult rat heart | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
CaSki | Human cervical carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
CHO K1 | Hamster ovary | Plasmid DNA | Lipofectamine® LTX | 15338-100 | 15338-500 | ![]() |
|
CHO-S | Hamster ovary | Protein Production | FreeStyle™ MAX Reagent | 16447-100 | 16447-500 | ![]() |
|
CMMT | Rhesus monkey mammary. tumor | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Cortical Neurons, E18 | Rat brain | Plasmid DNA | Lipofectamine® LTX | 15338-100 | 15338-500 | ![]() |
|
COS 7L | Monkey kidney fibroblast | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
COS 7L | Monkey kidney fibroblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
CV-1 | Primate kidney | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
CV-1 | Primate kidney | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
D3 | Mouse embryonic stem cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
D3 | Mouse embryonic stem cells | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Daoy | Human medulloblastoma cell line | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
DL-3 | Mouse T lymphoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
DLD-1 | Human colon adenocarcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
DT40 | Chicken Bursa | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
EOC | Human epithelial ovarian cancer | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Foreskin Fibroblast, Primary | Human foreskin | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
G - H | |||||||
Glial Cell | Mouse brain | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
GripTite™ 293 MSR | Human kidney | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HaCaT | Human immortalized keratinocytes | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HASMC | Human aortic smooth muscle cells | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HCT116 | Human colon carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HCT116 | Human colon carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HEK 293 | Human embryonic kidney | Co-transfect Plasmid DNA and siRNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HEK 293 | Human embryonic kidney | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HEK 293 | Human embryonic kidney | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HeLa S3 | Human cervical carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HeLa S3 | Human cervical carcinoma | Co-transfect Plasmid DNA and siRNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HeLa S3 | Human cervical carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Hepatocytes, Primary | Mouse liver | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HepG2 | Human liver carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HepG2 | Human liver carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Hippocampal Neurons | Rat brain | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Hippocampal Neurons | Rat brain | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HL60 | Human lymphoblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HMEC-1 | Human dermal microvascular endothelial cells | Plasmid DNA | Lipofectamine® LTX | 15338-100 | 15338-500 | ![]() |
|
HMVEC-L | Human lung microvascular endothelial cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HNSCC | Human head and neck squamous cell carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HT 29 | Human colon adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HT1080 | Human fibrosarcoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HT1080 | Human fibrosarcoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HT1376 | Human grade III transitional bladder cell carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Huh-7 | Human hepatocellular carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Huh-7 | Human hepatocellular carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Human Mesenchymal Stem Cells | Human bone marrow | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Human Mesenchymal Stem Cells | Human bone marrow | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
HUVEC | Human umbilical vein cord | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
HUVEC | Human umbilical vein cord | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
I - M | |||||||
IMR-90 | Human embryonic lung | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
IMR-90 | Human embryonic lung | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
INS-1 | Rat pancreatic {beta}-cell line | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
JEG-3 | Human choriocarcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Jurkat | Human T-cell leukemia | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
JygMC(A) | Mouse mammary carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
K562 | Human myelogenous leukemia | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
K562 | Human myelogenous leukemia | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
L929 | Mouse fibroblast cell line | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
LC-NCI-H23 | Non-small cell, epid. | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
LNCap | Human prostate cancer | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
LS180 | Human colon carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
LSMC | Human leiomyoma smooth muscle cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
M21 | Human melanoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
McA-RH7777 | Rat hepatoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MCF 7 | Human breast cancer | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MCF 7 | Human breast cancer | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
MCF 7 | Human breast cancer | Co-transfect Plasmid DNA and siRNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
MDA-MB-231 | Human breast adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
MDA-MB-435 | Human breast adenocarcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MDA-MB-435 | Human breast adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
MDCK | Madin-Darby canine kidney | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
MDCK | Madin-Darby canine kidney | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MDM | Human monocyte-derived macrophages | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
ME-180 | Human cervical carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MEF | Mouse embryonal fibroblast cells | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Mesangial Cells, Primary | Rat kidney | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
mIMCD3 | Murine inner medullary collecting duct | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MIN6 | Mouse pancreatic {beta}-cell line | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
MLT | Mouse Leydig tumor | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
Molt-4 | Human T lymphoblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
MRC-5 | Human lung | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
N - R | |||||||
N2a-APP | Mouse neuroblastoma N2a cells stably expressing human APP695 | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
N-38 | Mouse hypothalamic cell line | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NB | Human neuroblastoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NBS | Human Nijmegen breakage syndrome cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NCI-H460 | Human large cell lung cancer | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
NDHF | Neonatal dermal fibroblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Neonatal Rat Ventricular Cardiomyocyte | Rat heart muscle | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ||
Neural Progenitor Cells, Primary | Embryonic mouse brain | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Neuro2A | Mouse brain neuroblastoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Neuro2A | Mouse brain neuroblastoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NG108-15 | Hybrid of mouse neuroblastoma N18TG2 and rat glioma C6-BU-1 cells | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
NHFF | Human diploid somatic fibroblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
NIH 3T3 | Mouse embryonic fibroblast | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
NIH 3T3 | Mouse embryonic fibroblast | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NOR-10 (H-2b) | Mouse skeletal muscle cell line | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NRK | Rat normal kidney | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NT2 | Human teratocarcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
NTera2 | Human embryonic carcinoma stem cell | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
OLN-93, Primary | Rat brain glial cultures | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
OLP, Primary | Rat cerebra | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
P19 | Mouse embryonal carcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
P19 | Mouse embryonal carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
PC12 | Rat neuronal | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
PC3 | Human prostate adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Rat1 | Rat fibroblast cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
RAW264.7 | Mouse macrophage cells | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
RPC-C2A | Rat dental pulp cells | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
S - W | |||||||
S2-013 | Human pancreas | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Saos-2 | Human osteosarcoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Sf9 | Insect ovary | DNA | Cellfectin® | 10362-010 | ![]() |
||
SK-BR3 | Human breast carcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
SK-MEL-28 | Human malignant melanoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
SK-N-SH | Human neuroblastoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
SK-N-SH | Human neuroblastoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
SK-OV-3 | Human ovarian adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
SNB19 | Human glioblastoma cell line | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
SW1417 | Human colon adenocarcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
SW480 | Human colon adenocarcinoma | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
T47D | Human mammary adenocarcinoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
T98G | Human brain | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
THP-1 | Human monocytic leukemia | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
THP-1 | Human monocytic leukemia | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
U251 | Human glial | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
U2OS | Human osteosarcoma | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |
|
U87 | Human astrocyte | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
Vero | African green monkey kidney | Plasmid DNA | Lipofectamine® 3000 | L3000008 | L3000015 | ![]() |
|
WI-38(VA13) | Normal human fetal lung fibroblast | siRNA | Lipofectamine® RNAiMAX | 13778-075 | 13778-150 | ![]() |